Skip to Content
Merck
All Photos(1)

Key Documents

Z742620

Tube-O-CONCENTRATOR Concentration Device

0.2 ‑ 2.5 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123100
NACRES:
NA.56

feature

0.2 ‑ 2.5 mL

packaging

pkg of 5 ea

manufacturer/tradename

G-BioSciences 786-626

working volume

0.2—2.5 mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Include:
  • Concentrator Solution - 125 ml
  • Tube-O-DIALYZER, 1,000MWCO, Micro - sample volume up to 250 ul - 5 units
  • Micro Dialysis Cups - 5 units
  • Floats - 5 units
  • Storage Caps - 5 units

Tube-O-CONCENTRATOR is a versatile concentration device that utilizes a novel liquid polymer and our patented
Tube-O-CONCENTRATOR for the rapid concentration of dilute protein solutions with zero loss. The unique tube design ensures that 100% sample is recovered; simple place the entire device in a bench top centrifuge and spin for a few seconds.
The Tube-O-CONCENTRATOR solution is a liquid polymer that rapidly absorbs water through the dialysis membrane in the Tube-O-CONCENTRATOR cap, which retains all molecules with a molecular weight >1kDa.
Tube-O-CONCENTRATOR is available in two sizes for concentrating sample volumes of up to 250μl (Micro) or 2.5ml (Medi).

Legal Information

Tube-O-CONCENTRATOR is a trademark of Geno Technology, Inc.
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

149.0 °F

Flash Point(C)

65 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service