Skip to Content
Merck
All Photos(1)

Key Documents

Z729027

Hettich® bucket for rotor 1324

for ROTOFIX 32A and UNIVERSAL 320/320R, Bucket for Rotor 1324 (Z722790)

Synonym(s):

Hettich® centrifuge buckets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103910
NACRES:
NB.22

feature

Holds 1 x 50 mL tubes
Holds 4 x 2.6 - 15 mL tubes
Holds 5 x 5 - 7 mL tubes

manufacturer/tradename

Hettich 1398

technique(s)

centrifugation: suitable

compatibility

for use with Rotor 1324

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Bucket 1398 (Z729027) is not compatible with any lid

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service