Key Documents
Z720933
Nunc® Cell Factory accessories
Start-up Kit, sterile, 1/cs
Synonym(s):
cell harvesting system, closed cell culture system
About This Item
UNSPSC Code:
41161502
NACRES:
NB.22
Pricing and availability is not currently available.
sterility
sterile
feature
Starter kit includes: PC Connector, Airvent Filter, White Filter Adaptor Cap, Cover Caps, Tube Clamp and Silicone Tubing
packaging
case of 1
pack of 1
manufacturer/tradename
Nunc 170769
General description
Starter kit includes: PC Connector, Airvent Filter, White Filter Adaptor Cap, Cover Caps, Tube Clamp and Silicone Tubing
Other Notes
Cell Factory accessories for filling and emptying the cell factories with media
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)