Z326755
BRAND® BLAUBRAND® ETERNA volumetric flask
volume 50 mL, accuracy: 0.08 mL, neck joint: ST/NS 14/23
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
wide-neck
Joint
joint: ST/NS neck14/23
description
calibrated to contain (TC, EX)
sterility
non-sterile
feature
accuracy class A (DIN EN ISO 1042, calibrated to contain (TC, EX))
closure type PP Stopper
packaging
pack of 2
manufacturer/tradename
BRAND
volume measuring range
50 mL, accuracy: 0.08 mL
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
General description
Class A, conformity certified. Made from Boro 3.3, Supplied with a square grip PP stopper. Marks and inscriptions in ETERNA amber diffusion stain, particularly resistant to aggressive cleaning methods. TO DIN EN ISO 1042.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR36848 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service