Skip to Content
Merck
All Photos(5)

Key Documents

WH0008940M1

Sigma-Aldrich

Monoclonal Anti-TOP3B antibody produced in mouse

clone 4F11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-topoisomerase (DNA) III beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOP3B(8940)

Related Categories

General description

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus relaxing the supercoils and altering the topology of DNA. The enzyme interacts with DNA helicase SGS1 and plays a role in DNA recombination, cellular aging and maintenance of genome stability. Alternative splicing of the C-terminus of this gene results in three transcript variants which have distinct tissue specificity; however, not all variants have been fully described. (provided by RefSeq)

Immunogen

TOP3B (AAH02432, 1 a.a. ~ 862 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MKTVLMVAEKPSLAQSIAKILSRGSLSSHKGLNGACSVHEYTGTFAGQPVRFKMTSVCGHVMTLDFLGKYNKWDKVDPAELFSQAPTEKKEANPKLNMVKFLQVEGRGCDYIVLWLDCDKEGENICFEVLDAVLPVMNKAHGGEKTVFRARFSSITDTDICNAMACLGEPDHNEALSVDARQELDLRIGCAFTRFQTKYFQGKYGDLDSSLISFGPCQTPTLGFCVERHDKIQSFKPETYWVLQAKVNTDKDRSLLLDWDRVRVFDREIAQMFLNMTKLEKEAQVEATSRKEKAKQRPLALNTVEMLRVASSSLGMGPQHAMQTAERLYTQGYISYPRTETTHYPENFDLKGSLRQQANHPYWANTVKRLLAEGINRPRKGHDAGDHPPITPMKSATEAELGGDAWRLYEYITRHFIATVSHDCKYLQSTISFRIGPELFTCSGKTVLSPGFTEVMPWQSVPLEESLPTCQRGDAFPVGEVKMLEKQTNPPDYLTEAELITLMEKHGIGTDASIPVHINNICQRNYVTVESGRRLKPTNLGIVLVHGYYKIDAELVLPTIRSAVEKQLNLIAQGKADYRQVLGHTLDVFKRKFHYFVDSIAGMDELMEVSFSPLAATGKPLSRCGKCHRFMKYIQAKPSRLHCSHCDETYTLPQNGTIKLYKELRCPLDDFELVLWSSGSRGKSYPLCPYCYNHPPFRDMKKGMGCNECTHPSCQHSLSMLGIGQCVECESGVLVLDPTSGPKWKVACNKCNVVAHCFENAHRVRVSADTCSVCEAALLDVDFNKAKSPLPGDETQHMGCVFCDPVFQELVELKHAASCHPMHRGGPGRRQGRGRGRARRPPGKPNPRRPKDKMSALAAYFV

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongyi Xu et al.
Nature neuroscience, 16(9), 1238-1247 (2013-08-06)
Topoisomerases are crucial for solving DNA topological problems, but they have not been linked to RNA metabolism. Here we show that human topoisomerase 3β (Top3β) is an RNA topoisomerase that biochemically and genetically interacts with FMRP, a protein that is
Yuyoung Joo et al.
Nature communications, 11(1), 3143-3143 (2020-06-21)
Topoisomerase 3β (Top3β) is the only dual-activity topoisomerase in animals that can change topology for both DNA and RNA, and facilitate transcription on DNA and translation on mRNAs. Top3β mutations have been linked to schizophrenia, autism, epilepsy, and cognitive impairment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service