Skip to Content
Merck
All Photos(1)

Key Documents

V7005

Supelco

Liquid scintillation vials, glass

volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with Poly-Seal cone liner. (caps packed separately), pkg of 500 ea

Synonym(s):

Glass Scintillation Vials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32
Pricing and availability is not currently available.

material

glass vial

feature

closure type screw top vial
closure type white urea cap

packaging

pkg of 500 ea

vial size

28 mm × 61 mm , with cap

volume

20 mL

fitting

thread for 22-400

Looking for similar products? Visit Product Comparison Guide

General description

Wheaton 180 brand low-potassium glass yielding consistent low backgrounds and high UV transmission. White caps fit tightly and can be written on.

Application



  • The determination of the radioactivity of 14C samples adsorbed on glass vials by direct liquid scintillation counting.: This study, published in Analytical Biochemistry, explores the application of glass vials in direct liquid scintillation counting of 14C samples. The research highlights the efficiency and accuracy of using glass vials for radioactivity measurements, providing a reliable method for chemists involved in radioisotope analysis (Wigfield DC, 1974).


Packaging

Vials are shrink-wrapped in individual cell trays of 100.
Caps are packed in sacks of 100 in separate trays.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuo S Okubo et al.
Neuron, 107(5), 924-940 (2020-07-19)
Spatial maps in the brain are most accurate when they are linked to external sensory cues. Here, we show that the compass in the Drosophila brain is linked to the direction of the wind. Shifting the wind rightward rotates the

Questions

  1. What is a poly-seal cone liner in the cap of the V7005-500EA liquid scintillation vials, glass?

    1 answer
    1. The Poly Seal Cone Liner is made of polyethylene.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service