Skip to Content
Merck
All Photos(3)

Key Documents

T122

Sigma-Aldrich

Telenzepine dihydrochloride hydrate

solid, ≥98% (HPLC)

Synonym(s):

4,9-Dihydro-3-methyl-4-[(4-methyl-1-piperazinyl)acetyl]-10H-thieno[3,4-b][1,5]benzodiazepin-10-one dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N4O2S · 2HCl · xH2O
CAS Number:
Molecular Weight:
443.39 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: >10 mg/mL

SMILES string

O.Cl.Cl.CN1CCN(CC1)CC(=O)N2c3ccccc3NC(=O)c4csc(C)c24

InChI

1S/C19H22N4O2S.2ClH.H2O/c1-13-18-14(12-26-13)19(25)20-15-5-3-4-6-16(15)23(18)17(24)11-22-9-7-21(2)8-10-22;;;/h3-6,12H,7-11H2,1-2H3,(H,20,25);2*1H;1H2

InChI key

HZWCRPGARSMPNS-UHFFFAOYSA-N

Gene Information

human ... CHRM1(1128)

Application

Telenzepine dihydrochloride hydrate was used to study M1 muscarinic receptor-mediated signaling in juvenile rat hippocampus and rat urinary bladder.

Biochem/physiol Actions

Telenzepine is an analog of pirenzepine and inhibits the secretion of gastric acid. It is an antagonist to M1-receptor and targets the oxyntic gastric glands and the salivary glands.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G Coruzzi et al.
Archives internationales de pharmacodynamie et de therapie, 302, 232-241 (1989-11-01)
The new M1-receptor antagonist telenzepine has been studied for its antisecretory effect in different in vitro and in vivo experimental models in comparison with pirenzepine. Telenzepine was found to be from 3 to 10 times more potent than pirenzepine in
G Bertaccini et al.
Pharmacological research, 21(4), 339-352 (1989-07-01)
The control of gastric secretion may be obtained by means of several pharmacological compounds: histamine H2 receptor antagonists and anticholinergics are so far the most widely employed drugs in pharmacological experiments and in clinical practice. The H2 blockers are able
K Kawashima et al.
General pharmacology, 21(1), 17-21 (1990-01-01)
1. Effects of three muscarinic antagonists on electrically evoked ACh release and contractile response were investigated in longitudinal muscle strips of guinea-pig ileum suspended in an organ-bath and superfused with Krebs solution. ACh release was determined by a specific radioimmunoassay.
Turgut Emrah Bozkurt et al.
Pharmacological research, 59(5), 300-305 (2009-05-07)
We have investigated the muscarinic receptor subtype(s) mediating the release of urinary bladder-derived relaxant factor that is demonstrated by a coaxial bioassay system. Acetylcholine-induced relaxation of a precontracted anococcygeus muscle mounted within the bladder was considered as an evidence for
A M Rosati et al.
Brain research, 815(2), 185-191 (1999-01-08)
Binding studies were used to assess the changes in affinity and/or number of M1 muscarinic receptors in hippocampi from juvenile rats chronically deprived of NGF. NGF deprivation was obtained by implanting into right ventricle at postnatal day 2 (P2) hybrydoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service