SRP3318
C1 Inhibitor Peptide
≥95% (HPLC), lyophilized powder, human recombinant, expressed in CHO cells
Synonym(s):
C1inh, Plasma protease C1 inhibitor, Serpin G1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
Product Name
C1 Inhibitor human, recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
biological source
human
recombinant
expressed in CHO cells
Assay
≥95% (HPLC)
≥95% (SDS-PAGE)
form
lyophilized
potency
≤2.6 nM IC50
mol wt
52.8 kDa
packaging
pkg of 200 μg
impurities
endotoxin, tested
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... SERPING1(710)
General description
Plasma protease C1 inhibitor is a member of the serpin family of structurally related proteins. Recombinant Human C1 inhibitor is a highly glycosylated glycoprotein containing 478 amino acid residues (52.8kDa), corresponding to amino acids 23-500 of the C1 inhibitor precursor, and is fully functional in its ability to inhibit the C1 complex. Glycosylated C1 inhibitor migrates at an apparent molecular weight of approximately 80-90kDa by SDS PAGE analysis under reducing conditions.
Biochem/physiol Actions
Plasma protease C1 inhibitor is the primary regulator of the immune complement system. C1 inhibitor is a protease inhibitor that functions to inhibit the complement system in order to prevent over-activation or spontaneous activation. Inhibition is achieved by binding to and irreversibly inhibiting the C1r and C1s proteases of the C1 complex, which has the effect of shutting down all subsequent downstream events in the complement activation cascade. C1 inhibitor can also inhibit various other proteases, including Kallikrein, Factor XIa, and Factor XIIa. Deficiencies in C1 inhibitor are the primary cause of hereditary angioedema (HAE, hereditary angioneurotic edema), a disease characterized by edema in the respiratory and gastrointestinal tracts. In certain clinical situations, the direct administration of C1 inhibitor can be used to treat HAE and certain other conditions.
Physical form
Lyophilized from 10mM Sodium Phosphate, pH 7.5.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C1 Esterase Inhibitor (Human)
P & T : A Peer-Reviewed Journal for Formulary Management, 35, 2-3 (2010)
C1 inhibitor.
A E Davis et al.
Methods in enzymology, 223, 97-120 (1993-01-01)
Sayani Mukherjee et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(41), 7965-7979 (2020-09-06)
Microglia, a type of CNS immune cell, have been shown to contribute to ethanol-activated neuronal death of the stress regulatory proopiomelanocortin (POMC) neuron-producing β-endorphin peptides in the hypothalamus in a postnatal rat model of fetal alcohol spectrum disorders. We determined
C1-inhibitor deficiency and angioedema.
Carugati A
Molecular Immunology, 38(2-3), 161-173 (2001)
In vivo biosynthesis of endogenous and of human C1 inhibitor in transgenic mice: tissue distribution and colocalization of their expression.
Vinci G
Journal of Immunology, 169(10), 5948-5954 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service