SRP0551
Monkey (M. fascicularis) PD-1, FC Fusion
recombinant, expressed in HEK 293 cells
Synonym(s):
CD279, HPD-L, PD-1, PDCD1, Programmed cell death 1, SLEB2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.26
recombinant
expressed in HEK 293 cells
form
frozen liquid
mol wt
42.6 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
Monkey ... PDCD1 (100135775)
General description
Monkey secreted Programmed Cell Death 1 (PD-1)-Fc fusion protein, also known as CD279, PDCD1, and SLEB2, GenBank Accession No. NM_005018, a.a. 25-167, fused at the C-terminus to the Fc portion of human IgG1, expressed in a HEK293 cell expression system. MW = 42.6 kDa. Note: this protein runs at an apparent higher molecular weight due to glycosylation.
Physical form
Aqueous solution
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service