Skip to Content
Merck
All Photos(2)

Documents

SML3941

Sigma-Aldrich

DUB-IN-1

≥98% (HPLC)

Synonym(s):

9-Benzyloxyimino-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile, 9-Phenylmethoxyimino-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile, DUB Inhibitor 1, DUBs Inhibitor 1, DUBs-IN-1, Deubiquitinase Inhibitor 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H11N5O
CAS Number:
Molecular Weight:
337.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Potent and selective ubiquitin specific peptidase USP8 deubiquitinase (DUB) inhibitor.

DUB-IN-1 is a potent and selective ubiquitin specific peptidase USP8 deubiquitinase (DUB) inhibitor (IC50 = 850 nM/USP8, >100 µM/USP7). DUB-IN-1 inhibits the proliferation (200-800 nM for 24 or 48h; LN229, U87MG, and T98G cells), invasion (500-800 nM for 48 or 96h; LN229 and T98G cells), and stemness (500-800 nM for 7 or 14 days; LN229 and T98G cells) in GBM cultures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beibei Sha et al.
Cell biology and toxicology, 39(5), 2011-2032 (2022-01-14)
Increasing evidence suggests that targeting ubiquitin-specific peptidase 8 (USP8) serves as an attractive anti-cancer strategy. However, the role of USP8 inhibitor, DUB-IN-1, in esophageal squamous cell carcinoma (ESCC) cells still needs to be explored. Here, immunohistochemistry was employed to examine
Matteo Colombo et al.
ChemMedChem, 5(4), 552-558 (2010-02-27)
High-throughput screening highlighted 9-oxo-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile (1) as an active inhibitor of ubiquitin-specific proteases (USPs), a family of hydrolytic enzymes involved in the removal of ubiquitin from protein substrates. The chemical behavior of compound 1 was examined. Moreover, the synthesis and in
Yu Long et al.
Open life sciences, 18(1), 20220562-20220562 (2023-02-24)
Glioblastoma (GBM) is a malignant brain tumor. The purpose of this study is to estimate the potential effects and underlying mechanisms of a ubiquitin-specific protease 8 (USP8) small-molecule inhibitor on the phenotypic characteristics of GBM cells. The growth, migration, invasion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service