Skip to Content
Merck
All Photos(2)

Key Documents

SML3802

Sigma-Aldrich

ARN-3236

≥98% (HPLC)

Synonym(s):

3-(2,4-Dimethoxyphenyl)-4-(3-thienyl)-1H-pyrrolo[2,3-b]pyridine, 3-(2,4-Dimethoxyphenyl)-4-(thiophen-3-yl)-1H-pyrrolo[2,3-b]pyridin, ARN 3236, ARN3236

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16N2O2S
CAS Number:
Molecular Weight:
336.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1cc(cc1)c2c3c([nH]cc3c4c(cc(cc4)OC)OC)ncc2

InChI

1S/C19H16N2O2S/c1-22-13-3-4-15(17(9-13)23-2)16-10-21-19-18(16)14(5-7-20-19)12-6-8-24-11-12/h3-11H,1-2H3,(H,20,21)

InChI key

WEHOIIGXTMKVRG-UHFFFAOYSA-N

Biochem/physiol Actions

ARN-3236 is an orally active, ATP-competitive, SIK2-selective salt-inducible kinase inhibitor (IC50 = 21.63 nM/SIK1, <1 nM/SIK2, 6.63 nM/SIK3) that suppresses cellular SIK2 activity (by 58% at 1 μM; SKOv3-SIK2 cells, 48 h) with high selectivity over >456 kinases. ARN-3236 effectively suppresses SIK2-dependent growth of ovarian cancer cultures (IC50 = 0.8-2.6 μM in 10 lines, 72 h) and in murine xenograft models in vivo (30, 60, 100 mg/kg/day p.o.). In addition, ARN-3236 sensitizes ovarian cancer cells to paclitaxel in vitro and in vivo. ARN-3236 completes the SIK1-selective HG-9-91-01 (SIK1/2/3 IC50 = 0.6/6.6/9.6 nM) in probing SIKs-mediated cellular signaling events.
Orally active, ATP-competitive, highly potent and selective SIK2 inhibitor with anti-ovarian cancer efficacy in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhua Zhou et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(8), 1945-1954 (2016-09-30)
Purpose: Salt-inducible kinase 2 (SIK2) is a centrosome kinase required for mitotic spindle formation and a potential target for ovarian cancer therapy. Here, we examine the effects of a novel small-molecule SIK2 inhibitor, ARN-3236, on sensitivity to paclitaxel in ovarian
Tissue viability. Measuring the pressure.
M Clark
Nursing times, 84(25), 72-75 (1988-06-22)
Venipuncture: simple and aafe.
C E Murphy
Dental survey, 47(1), 31-33 (1971-01-01)
Yue Liu et al.
Frontiers in pharmacology, 11, 624429-624429 (2021-02-02)
Depression is a widespread chronic medical illness affecting thoughts, mood, and physical health. However, the limited and delayed therapeutic efficacy of monoaminergic drugs has led to intensive research efforts to develop novel antidepressants. ARN-3236 is the first potent and selective
Kimberly E Maxfield et al.
Molecular and cellular biology, 36(24), 3048-3057 (2016-10-05)
Triple-negative breast cancer (TNBC) is a highly heterogeneous disease with multiple, distinct molecular subtypes that exhibit unique transcriptional programs and clinical progression trajectories. Despite knowledge of the molecular heterogeneity of the disease, most patients are limited to generic, indiscriminate treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service