Skip to Content
Merck
All Photos(1)

Key Documents

SML3472

Sigma-Aldrich

ATR-002

≥98% (HPLC)

Synonym(s):

Benzoic acid, 2-[(2-chloro-4-iodophenyl)amino]-3,4-difluoro- (9CI, ACI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H7ClF2INO2
CAS Number:
Molecular Weight:
409.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(C(F)=C1NC2=CC=C(C=C2Cl)I)F)O

InChI

1S/C13H7ClF2INO2/c14-8-5-6(17)1-4-10(8)18-12-7(13(19)20)2-3-9(15)11(12)16/h1-5,18H,(H,19,20)

InChI key

XCNBGWKQXRQKSA-UHFFFAOYSA-N

Biochem/physiol Actions

ATR-002, a derivate of CI-1040, is a cell penetrant, potent and specific inhibitor of the Raf/MEK/ERK pathway. ATR-002 exhibits strong anti-influenza and anti-SARS-CoV-2 activity. It impairs the influenzas and SARS-CoV-2 viruses induced expression of pro-inflammatory cytokines.
cell penetrant, potent and specific inhibitor of the Raf/MEK/ERK pathway; exhibits strong anti-influenza and anti-SARS-CoV-2 activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Schreiber et al.
Cellular and molecular life sciences : CMLS, 79(1), 65-65 (2022-01-12)
Coronavirus disease 2019 (COVID-19), the illness caused by a novel coronavirus now called severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has led to more than 260 million confirmed infections and 5 million deaths to date. While vaccination is a powerful
Martin Laure et al.
Antiviral research, 178, 104806-104806 (2020-04-19)
Antiviral therapies against influenza are required, especially for high-risk patients, severe influenza and in case of highly pathogenic influenza virus (IV) strains. However, currently, licensed drugs that target the virus directly are not very effective and often lead to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service