Skip to Content
Merck
All Photos(1)

Key Documents

SML2813

Sigma-Aldrich

UNC6934

≥98% (HPLC)

Synonym(s):

3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-7-carboxylic acid cyclopropyl-[4-(pyrimidin-4-ylcarbamoyl)-benzyl]-amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21N5O4
CAS Number:
Molecular Weight:
443.45
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(CN(C2CC2)C(C3=CC4=C(NC(CO4)=O)C=C3)=O)C=C1)NC5=CC=NC=N5

InChI

1S/C24H21N5O4/c30-22-13-33-20-11-17(5-8-19(20)27-22)24(32)29(18-6-7-18)12-15-1-3-16(4-2-15)23(31)28-21-9-10-25-14-26-21/h1-5,8-11,14,18H,6-7,12-13H2,(H,27,30)(H,25,26,28,31)

InChI key

KOZGEDUWAQFVAV-UHFFFAOYSA-N

Biochem/physiol Actions

SGC probe under development. UNC6934 is a potent and selective antagonist of histone-lysine N-methyltransferase NSD2-PWWP1 domain (Nuclear Receptor Binding SET Domain Protein 2 PWWP1 domain). UNC6934 disrupts the NSD2-PWWP1 interaction with H3K36me2 nucleosomes in U2OS cells.
potent and selective antagonist of NSD2-PWWP1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service