Skip to Content
Merck
All Photos(1)

Key Documents

SML2399

Sigma-Aldrich

ML327

≥98% (HPLC)

Synonym(s):

1,2-Dihydro-2-oxo-N-[3-[[(5-phenyl-3-isoxazolyl)carbonyl]amino]propyl]-3-pyridinecarboxamide, CID 60167648

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N4O4
CAS Number:
Molecular Weight:
366.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1[c](c(ccc1)C(=O)NCCCNC(=O)c2n[o]c(c2)c3ccccc3)=O

InChI key

NNNDNXLMQAPQQQ-UHFFFAOYSA-N

Biochem/physiol Actions

ML327 is a regulator of E-cadherin transcription that restores E-cadherin expression in cancer cell lines and partially reverses Epithelial-to-Mesenchymal Transition (EMT). It is a potent inducer of mesenchymal-to-epithelial transition (MET) in epithelial cancers. Apparently ML327 blocks MYC expression in neuroblastomas. It induces apoptosis in cancer cells and sensitizes Ewing sarcoma cells to TRAIL.
regulator of E-cadherin transcription that restores E-cadherin expression in cancer cell lines; inducer of mesenchymal-to-epithelial transition (MET)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanbing An et al.
Oncotarget, 6(26), 22934-22948 (2015-06-18)
Transcriptional repression of E-cadherin is a hallmark of Epithelial-to-Mesenchymal Transition (EMT) and is associated with cancer cell invasion and metastasis. Understanding the mechanisms underlying E-cadherin repression during EMT may provide insights into the development of novel targeted therapeutics for cancer.
Eric J Rellinger et al.
Biochemical and biophysical research communications, 491(2), 463-468 (2017-07-19)
Ewing sarcomas are rare mesenchymal-derived bone and soft tissue tumors in children. Afflicted children with distant metastases have poor survival despite aggressive therapeutics. Epithelial-to-mesenchymal transition in epithelial carcinomas is associated with loss of E-cadherin and resistance to apoptosis. ML327 is
Chandrasekhar Padmanabhan et al.
Oncotarget, 8(60), 101072-101086 (2017-12-20)
Epithelial cancers (carcinomas) comprise the top four causes of cancer-related deaths in the United States. While overall survival has been steadily improving, therapy-resistant disease continues to present a major therapeutic challenge. Carcinomas often exploit the normal developmental program, epithelial-to-mesenchymal transition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service