Skip to Content
Merck
All Photos(1)

Key Documents

SML2343

Sigma-Aldrich

SGC3027

≥98 mg/mL (HPLC)

Synonym(s):

SGC3027, 5′-S-[4-[(N-(3-(2,4,5-trimethyl-3,6-dioxo-cyclohexa-1,4-dienyl)-3-methyl)butyryl)-(N-(4′-chloro-biphenyl-3-ylmethyl)amino]butyl]-5′-thioadenosine, N-(4-((((2S,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)thio)butyl)-N-((4′-chloro-[1,1′-biphenyl]-3-yl)methyl)-3-methyl-3-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)butanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H47ClN6O6S
CAS Number:
Molecular Weight:
787.37
UNSPSC Code:
12352200
NACRES:
NA.77

form

powder

concentration

≥98 mg/mL (HPLC)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC(C=C1)=CC=C1C2=CC(CN(C(CC(C)(C)C(C(C(C)=C3C)=O)=C(C)C3=O)=O)CCCCSC[C@H]4O[C@@H](N(C=N5)C6=C5C(N)=NC=N6)[C@H](O)[C@@H]4O)=CC=C2

InChI

1S/C41H47ClN6O6S/c1-23-24(2)35(51)32(25(3)34(23)50)41(4,5)18-31(49)47(19-26-9-8-10-28(17-26)27-11-13-29(42)14-12-27)15-6-7-16-55-20-30-36(52)37(53)40(54-30)48-22-46-33-38(43)44-21-45-39(33)48/h8-14,17,21-22,30,36-37,40,52-53H,6-7,15-16,18-20H2,1-5H3,(H2,4

InChI key

MLJVGAYSVYMPSB-MSUKGTQXSA-N

Biochem/physiol Actions

SGC3027 is a selective and cell-active inhibitor of protein arginine N-methyltransferase (PRMT7) It is a pro-drug of the active component, SGC8158, which is produced in the cell by reductases. SGC8158 was shown to inhibit PRMT7 with an IC50 value < 2.5 nM for methylation of H2B (23-37) and greater than 40-fold selectivity over other histone methyltransferases and non-epigenetic targets. SGC3027 inhibited the methylation of HSP70 In cellular assays using C2C12 cells with an IC50 value of 1.3 microM. The closely related SGC3027N is significantly less active in the cellular assay, and is a control compound for cellular studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service