Skip to Content
Merck

SML2273

KUNB31

≥98% (HPLC)

Synonym(s):

(4-Hydroxy-7-isopropylbenzo[d]isoxazol-5-yl)(isoindolin-2-yl)methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18N2O3
CAS Number:
Molecular Weight:
322.36
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N3(Cc4c(cccc4)C3)C(=O)c1c(c2c([o]nc2)c(c1)C(C)C)O

InChI key

MFZKTINXWPQHBG-UHFFFAOYSA-N

Biochem/physiol Actions

Isoform-selective inhibitor of Hsp90β
KUNB31 is an isoform-selective inhibitor of Hsp90β with an IC50 value of 180 nM and > 50-fold selectivity over closely related isoforms Hsp90α and GRP94.Hsp90 inhibitors currently in clinical trials are all pan-inhibitors with multiple side effects including induction of the pro-survival heat shock response. KUNB31 was shown to induce the degradation of Hsp90β-dependent client proteins, but not those of Hsp90α and GRP94, and did not induce production of Hsp90. It showed anti-proliferative activity against NCI H23 (non-small cell lung cancer), UC3 (bladder cancer), and HT-29 (colon adenocarcinoma) cells with low micromolar IC50 values, while requiring more than 100 μM against non-cancerous human embryonic kidney HEK-293 cells.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library