Skip to Content
Merck
All Photos(1)

Documents

SML2098

Sigma-Aldrich

KINK-1 hydrochloride

≥98% (HPLC)

Synonym(s):

7-[2-(Cyclopropylmethoxy)-6-hydroxyphenyl]-1,4-dihydro-5-(3S)-3-piperidinyl-2H-pyrido[2,3-d][1,3]oxazin-2-one hydrochloride, 7-[2-(Cyclopropylmethoxy)-6-hydroxyphenyl]-5-[(3S)-3-piperidinyl]-1,4-dihydro-2H-pyrido[2,3-d][1,3]oxazin-2-one hydrochloride, Bay 65-1942, CmpdA, Compound A, Kinase Inhibitor of NF-ΚB-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25N3O4 · HCl
CAS Number:
Molecular Weight:
431.91
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -19 to -24°, c = 0.2 in DMF

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

OC1=CC=CC(OCC2CC2)=C1C3=NC(NC(OC4)=O)=C4C([C@@H]5CCCNC5)=C3.Cl

InChI

1S/C22H25N3O4.ClH/c26-18-4-1-5-19(28-11-13-6-7-13)20(18)17-9-15(14-3-2-8-23-10-14)16-12-29-22(27)25-21(16)24-17;/h1,4-5,9,13-14,23,26H,2-3,6-8,10-12H2,(H,24,25,27);1H/t14-;/m1./s1

InChI key

XZTOAEZYOFWVHB-PFEQFJNWSA-N

Biochem/physiol Actions

KINK-1 (Kinase Inhibitor of NF-ΚB-1; Bay 65-1942; CmpdA; Compound A) is an ATP-competitive, IKKβ-selective IΚB kinase (IKK) inhibitor (Ki against ATP = 2 nM/IKKβ and 135 nM/IKKα) that potently inhibits IKKβ-catalyzed GST-IΚBα(1–54) phosphorylation (IC50 = 4 nM, 200 nM ATP) without affecting (IC50 >10μM) IKK3 or 27 other kinases, lipases, phosphatases, caspases, and MMPs. KINK-1 effectively inhibits NF-ΚB activation and cytokines production in various human and murine cultures (IC50 from 18 to 502 nM) and is efficacious against LPS-induced plasma TNF-α production in mice and rats in vivo (EC50 = 9.1 and 6.6 mg/kg p.o., repectively) with good pharmacokinetic profile and oral availability. KINK-1 is also shown to effectively inhibit OVA-induced lung inflammation in a rat model of asthma (ED50 <0.3 mg/kg p.o.) without adverse effects to the animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth A Fowler et al.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation, 23(4), 569-580 (2017-02-06)
Idiopathic pneumonia syndrome (IPS) is a noninfectious inflammatory disorder of the lungs that occurs most often after fully myeloablative allogeneic hematopoietic stem cell transplantation (HSCT). IPS can be severe and is associated with high 1-year mortality rates despite existing therapies.
Yanting Zhang et al.
Molecular cancer therapeutics, 15(7), 1504-1514 (2016-05-20)
NF-κB plays an important role in many types of cancer, including prostate cancer, but the role of the upstream kinase of NF-κB, IKKβ, in prostate cancer has neither been fully documented nor are there any effective IKKβ inhibitors used in
Eyra Marien et al.
Oncotarget, 7(11), 12582-12597 (2016-02-11)
Lung cancer is the leading cause of cancer death. Beyond first line treatment, few therapeutic options are available, particularly for squamous cell carcinoma (SCC). Here, we have explored the phospholipidomes of 30 human SCCs and found that they almost invariably
Zhipeng Li et al.
Oncotarget, 7(22), 31892-31906 (2016-02-20)
The overexpression or mutation of epidermal growth factor receptor (EGFR) has been associated with a number of cancers, including head and neck squamous cell carcinoma (HNSCC). Increasing evidence indicates that both the phosphatidylinositol-3-kinase (PI3K)-Akt-mammalian target of Rapamycin (mTOR) and the
Nancy C Moss et al.
American journal of physiology. Heart and circulatory physiology, 293(4), H2248-H2253 (2007-08-07)
Despite years of experimental and clinical research, myocardial ischemia-reperfusion (IR) remains an important cause of cardiac morbidity and mortality. The transcription factor nuclear factor-kappaB (NF-kappaB) has been implicated as a key mediator of reperfusion injury. Activation of NF-kappaB is dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service