Skip to Content
Merck
All Photos(1)

Key Documents

SML2082

Sigma-Aldrich

Scrambled 10Panx1 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

10PanxSCR TFA, FSVYWAQADR TFA, L-phenylalanyl-L-seryl-L-valyl-L-tyrosyl-L-tryptophyl-L-alanyl-L-glutaminyl-L-alanyl-L-a-aspartyl-L-arginine TFA, Phe-Ser-Val-Tyr-Trp-Ala-Gln-Ala-Asp-Arg TFA, SCPanx, TFA, Scrambled 10Panx TFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H79N15O16 · xC2HF3O2
CAS Number:
Molecular Weight:
1242.34 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

film (Colorless)

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

[nH]1c2c(c(c1)CC(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(N)Cc4ccccc4)CO)C(C)C)Cc3ccc(cc3)O)C(=O)NC(C)C(=O)NC(CCC(=O)N)C(=O)NC(C)C(=O)NC(CC(=O)O)C(=O)NC(CCCN=C(N)N)C(=O)O)cccc2

InChI

1S/C58H79N15O16/c1-29(2)47(73-55(86)44(28-74)72-50(81)37(59)23-32-11-6-5-7-12-32)56(87)71-41(24-33-16-18-35(75)19-17-33)53(84)70-42(25-34-27-64-38-14-9-8-13-36(34)38)52(83)66-30(3)48(79)67-39(20-21-45(60)76)51(82)65-31(4)49(80)69-43(26-46(77)78)54(85)68-40(57(88)89)15-10-22-63-58(61)62/h5-9,11-14,16-19,27,29-31,37,39-44,47,64,74-75H,10,15,20-26,28,59H2,1-4H3,(H2,60,76)(H,65,82)(H,66,83)(H,67,79)(H,68,85)(H,69,80)(H,70,84)(H,71,87)(H,72,81)(H,73,86)(H,77,78)(H,88,89)(H4,61,62,63)

InChI key

VNQGVHMGXBRHRA-UHFFFAOYSA-N

Biochem/physiol Actions

SCPanx (10PanxSCR) corresponds to the scrambled version of the pannexin-1 (panx1) inhibitor peptide 10panx1 and serves as a negative control peptide in pannexin-1 inhibitory studies involving 10panx1 treatment both in vitro and in vivo.
Scrambled control peptide for the reversible pannexin-1 (panx1) channel blocker 10Panx1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul W Furlow et al.
Nature cell biology, 17(7), 943-952 (2015-06-23)
During metastatic progression, circulating cancer cells become lodged within the microvasculature of end organs, where most die from mechanical deformation. Although this phenomenon was first described over a half-century ago, the mechanisms enabling certain cells to survive this metastasis-suppressive barrier
Juan Mauricio Garré et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(17), 4785-4801 (2016-04-29)
We show here that the growth factor FGF-1 is proinflammatory in the spinal cord and explore the inflammatory mechanisms. FGF-1 applied to rat spinal astrocytes in culture initiates calcium signaling and induces secretion of ATP that within minutes increases membrane
Nicholas L Weilinger et al.
Nature neuroscience, 19(3), 432-442 (2016-02-09)
Overactivation of neuronal N-methyl-D-aspartate receptors (NMDARs) causes excitotoxicity and is necessary for neuronal death. In the classical view, these ligand-gated Ca(2+)-permeable ionotropic receptors require co-agonists and membrane depolarization for activation. We report that NMDARs signal during ligand binding without activation
Roger J Thompson et al.
Science (New York, N.Y.), 322(5907), 1555-1559 (2008-12-06)
Pannexin-1 (Px1) is expressed at postsynaptic sites in pyramidal neurons, suggesting that these hemichannels contribute to dendritic signals associated with synaptic function. We found that, in pyramidal neurons, N-methyl-d-aspartate receptor (NMDAR) activation induced a secondary prolonged current and dye flux
C Yi et al.
Cell death and differentiation, 23(10), 1691-1701 (2016-07-09)
In Alzheimer's disease (AD), astrocyte properties are modified but their involvement in this pathology is only beginning to be appreciated. The expression of connexins, proteins forming gap junction channels and hemichannels, is increased in astrocytes contacting amyloid plaques in brains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service