Skip to Content
Merck
All Photos(1)

Key Documents

SML2063

Sigma-Aldrich

Davunetide trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

AL-108, Asn-Ala-Pro-Val-Ser-Ile-Pro-Gln, L-Asparaginyl-L-alanyl-L-prolyl-L-valyl-L-seryl-L-isoleucyl-L-prolyl-L-Glutamine, NAP, NAP peptide, NAPVSIPQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60N10O12 · xC2HF3O2
CAS Number:
Molecular Weight:
824.92 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

composition

Peptide content, ≥60%

color

white to beige

shipped in

wet ice

storage temp.

−20°C

SMILES string

N2([C@@H](CCC2)C(=O)N[C@@H](CCC(=O)N)C(=O)O)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H]1N(CCC1)C(=O)[C@@H](NC(=O)[C@@H](N)CC(=O)N)C)C(C)C)CO)[C@H](CC)C

InChI

1S/C36H60N10O12/c1-6-18(4)28(35(56)46-14-8-9-23(46)31(52)41-21(36(57)58)11-12-25(38)48)44-30(51)22(16-47)42-33(54)27(17(2)3)43-32(53)24-10-7-13-45(24)34(55)19(5)40-29(50)20(37)15-26(39)49/h17-24,27-28,47H,6-16,37H2,1-5H3,(H2,38,48)(H2,39,49)(H,40,50)(H,41,52)(H,42,54)(H,43,53)(H,44,51)(H,57,58)/t18-,19-,20-,21-,22-,23-,24-,27-,28-/m0/s1

InChI key

DWLTUUXCVGVRAV-XWRHUKJGSA-N

Biochem/physiol Actions

Davunetide (NAPVSIPQ; NAP) is an eight amino acid peptide identified as the active component of activity-dependent neuroprotective protein (ADNP). It has been found to be neuro-protective at femtomolar concentrations in vitro against the neural toxicity of a wide range of compounds. Davunetide has been investigated for Parkinson′s disease and schizophrenia and in clinical trials for Alzheimner′s Disease, targeting microtubule-tau Interaction.
Neuroprotective peptide; the neuronal tubulin-preferring agent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Zhang et al.
Physiology & behavior, 174, 67-73 (2017-03-05)
Memory loss and cognition decline are the main clinical manifestations of Alzheimer's disease (AD). Amyloid β protein (Aβ) aggregated in the brain is one of the key pathological characteristics of AD and responsible for the deficits in learning and memory.
Agata Grazia D'Amico et al.
Journal of molecular neuroscience : MN, 61(2), 256-266 (2016-12-03)
The retinal microvascular damage is a complication of diabetic retinopathy (DR). Hyperglycemia and hypoxia are responsible of aberrant vessel's proliferation. The cellular response to hypoxia is mediated through activation of hypoxia-inducible factors (HIFs). Among these, HIF-1α modulates expression of its
T Q Melo et al.
Experimental brain research, 235(3), 731-742 (2016-11-21)
The formation of oligomers and aggregates of overexpressed or mutant α-synuclein play a role in the degeneration of dopaminergic neurons in Parkinson's disease by causing dysfunction of mitochondria, reflected in their disturbed mobility and production of ROS. The mode of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service