Skip to Content
Merck
All Photos(1)

Key Documents

SML1356

Sigma-Aldrich

Sephin1

≥95% (HPLC)

Synonym(s):

(E)-2-(2-Chlorobenzylidene)hydrazinecarboximidamide, Sephin 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9ClN4
CAS Number:
Molecular Weight:
196.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC(N/N=C/C1=CC=CC=C1Cl)=N

InChI

1S/C8H9ClN4/c9-7-4-2-1-3-6(7)5-12-13-8(10)11/h1-5H,(H4,10,11,13)/b12-5+

InChI key

PDWJALXSRRSUHR-LFYBBSHMSA-N

Biochem/physiol Actions

In vivo studies using mice models show that sephin1 suppresses neurodegeneration, by preventing eIF2α (eukaryotic initiation factor 2) dephosphorylation.
Sephin1 is a selective inhibitor of a holophosphatase. It is a guanabez derivative that binds to and inhibits a regulatory subunit of the stress-induced protein phosphatase 1 (PPP1R15A), but not the constitutive PPP1R15B, and lacks α2-adrenergic activity. Phosphorylation of eIF2α, α subunit of eukaryotic translation initiation factor 2, reduces protein synthesis and prevents the accumulation of misfolded protein in the endoplasmic reticulum (ER). PPP1R15A recruits the serine/threonine-protein phosphatase PP1 to dephosphorylate eIF2α, so inhibiting PPP1R15A activity prolongs the phosphorylation of eIF2α and aids in its prevention of the accumulation of misfolded protein. In vitro Sephin1 protected cells from lethal protein misfolding and cytotoxic ER stress. In vivo sephin1 prevented two unrelated protein misfolding diseases in mice (Charchot-Marie-Tooth 1B and ALS).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PPP1R15A-mediated dephosphorylation of eIF2α is unaffected by Sephin1 or Guanabenz.
Crespillo-Casado A, et al.
eLife, 6, e26109-e26109 (2017)
Khang Nguyen et al.
Molecular metabolism, 83, 101921-101921 (2024-03-26)
Identification of new mechanisms mediating insulin sensitivity is important to allow validation of corresponding therapeutic targets. In this study, we first used a cellular model of skeletal muscle cell iron overload and found that endoplasmic reticulum (ER) stress and insulin
Nadejda Capatina et al.
The Journal of physiology, 599(17), 4153-4181 (2021-07-17)
Endoplasmic reticulum (ER) stress promotes placental dysmorphogenesis and is associated with poor pregnancy outcomes. We show that unfolded protein response signalling pathways located in the ER drive differentiation of mouse trophoblast stem cells into trophoblast subtypes involved in development of
Juliette Humeau et al.
Cell death & disease, 11(6), 433-433 (2020-06-10)
The integrated stress response is characterized by the phosphorylation of eukaryotic initiation factor-2α (eIF2α) on serine 51 by one out of four specific kinases (EIF2AK1 to 4). Here we provide three series of evidence suggesting that macroautophagy (to which we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service