Skip to Content
Merck
All Photos(1)

Key Documents

SML0975

Sigma-Aldrich

FLI-06

≥98% (HPLC)

Synonym(s):

1,4,5,6,7,8-Hexahydro-2,7,7-trimethyl-4-(4-nitrophenyl)-5-oxo-3-quinolinecarboxylic acid cyclohexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N2O5
CAS Number:
Molecular Weight:
438.52
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)C2C3=C(NC(=C2C(=O)OC4CCCCC4)C)CC(CC3=O)(C)C

InChI

1S/C25H30N2O5/c1-15-21(24(29)32-18-7-5-4-6-8-18)22(16-9-11-17(12-10-16)27(30)31)23-19(26-15)13-25(2,3)14-20(23)28/h9-12,18,22,26H,4-8,13-14H2,1-3H3

InChI key

SWWVFYHSSOWZMF-UHFFFAOYSA-N

Biochem/physiol Actions

FLI-06 is an inhibitor of Notch signaling. FLI-06 acts upstream of α-secretase and β-secretase cleavage, disrupting intracellular trafficking and processing of the Notch signaling pathway and inhibiting general secretion at a step before exit from the endoplasmic reticulum (ER). This inhibition is accompanied by a tubule-to-sheet morphological transition of the ER. FLI-06 does not act on the cytoskeleton, but causes a complete disruption of the Golgi in a manner different from that of Brefeldin A or Golgicide A. FLI-06 is the first compound acting that early in the secretory pathway.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yan Luan et al.
International journal of molecular sciences, 24(12) (2023-06-28)
Neural stem cells (NSCs) persist in the subgranular zone (SGZ) throughout the lifespan and hold immense potential for the repair and regeneration of the central nervous system, including hippocampal-related diseases. Several studies have demonstrated that cellular communication network protein 3
Rony Panarsky et al.
Cancer & metabolism, 8, 7-7 (2020-08-11)
The loss-of-function mutation of fumarate hydratase (FH) is a driver of hereditary leiomyomatosis and renal cell carcinoma (HLRCC). Fumarate accumulation results in activation of stress-related mechanisms leading to upregulation of cell survival-related genes. To better understand how cells compensate for
Anna-Marie Finger et al.
Science advances, 7(30) (2021-07-25)
Coupling between cell-autonomous circadian oscillators is crucial to prevent desynchronization of cellular networks and disruption of circadian tissue functions. While neuronal oscillators within the mammalian central clock, the suprachiasmatic nucleus, couple intercellularly, coupling among peripheral oscillators is controversial and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service