Skip to Content
Merck
All Photos(1)

Documents

SMB00701

Sigma-Aldrich

Sesamolin

≥97%, from Sesamum indicum (sesame)

Synonym(s):

5-[(1S,3aR,4R,6aR)-4-(1,3-Benzodioxol-5-yloxy)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O7
CAS Number:
Molecular Weight:
370.35
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

Sesamum indicum (sesame)

Quality Level

Assay

≥97%

form

powder

solubility

DMSO: 10 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[H][C@@]12[C@@H](C(C=C3)=CC4=C3OCO4)OC[C@]1([H])[C@@H](OC5=CC6=C(C=C5)OCO6)OC2

InChI

1S/C20H18O7/c1-3-15-17(25-9-23-15)5-11(1)19-13-7-22-20(14(13)8-21-19)27-12-2-4-16-18(6-12)26-10-24-16/h1-6,13-14,19-20H,7-10H2/t13-,14-,19+,20+/m0/s1

InChI key

ZZMNWJVJUKMZJY-AFHBHXEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sesamolin is a furofuran lignan extracted from Sesamum indicum L. (sesame) seeds. Its solubility is very less in water. The chemical structure of sesamolin comprises two phenylpropanoids with their propyl side attached to a central carbon. It is reported in many other Sesamum species.

Biochem/physiol Actions

It also exerts antimicrobial, anti-melanogenesis, and neuroprotective effects.
Sesamolin is reported to have beneficial physiological properties such as antioxidant, anti-mutagenic, anti-aging and anti-inflammatory activities. It was also found to inhibit lipid peroxidation in rat liver and kidney. Sesamolin enhances NK cell lysis activity by increasing the expression of NKG2D ligands on Burkitt′s lymphoma cells, presenting a potential immunotherapeutic value for cancer treatment. In addition, it was demonstrated that sesamolin inhibits β-secretase, presenting an important role for prevention of dementia by daily consumption.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Lind et al.
Acta orthopaedica Belgica, 58(1), 81-83 (1992-01-01)
The intra-articular pressures in the glenohumeral joint in different positions were measured in 12 cadaveric shoulders. The lowest pressure was found in abduction combined with traction. A significant rise in pressure was seen in positions combining flexion or abduction with
Reny Rosalina et al.
Molecules (Basel, Switzerland), 26(19) (2021-10-14)
Sesame seeds are rich in lignan content and have been well-known for their health benefits. Unlike the other sesame lignan compounds (i.e., sesamin and sesamol), the study of the pharmacological activity of sesamolin has not been explored widely. This review
M H Kang et al.
The Journal of nutrition, 128(6), 1018-1022 (1998-06-18)
Although the sesame lignans, sesaminol and sesamolinol, have been shown to possess antioxidative activity, less is known about the metabolism and antioxidative properties of sesamolin, a major constituent of sesame oil. To determine the ability of sesamolin to act as
Afaf Kamal-Eldin et al.
Recent patents on food, nutrition & agriculture, 3(1), 17-29 (2010-12-01)
Sesame seed contains very high levels (up to 2.5%) of furofuran lignans with beneficial physiological activities, mainly sesamin, sesamolin, and sesaminol glucosides. Reported activities of sesame seed lignans include inter alias modulation of fatty acid metabolism, inhibition of cholesterol absorption
β-secretase inhibitors from sesame seeds and turmeric rhizomes
Murata K, et al.
Planta Medica, 82, S1-S381 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service