Skip to Content
Merck
All Photos(1)

Key Documents

SMB00547

Sigma-Aldrich

Quercetin 3,4′-diglucoside

≥85% (LC/MS-UV)

Synonym(s):

Quercetin 3,4′-O-diglucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O17
CAS Number:
Molecular Weight:
626.52
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥85% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1=CC(O)=C2C(OC(C3=CC(O)=C(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)C=C3)=C(O[C@H]5[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O5)C2=O)=C1

InChI

1S/C27H30O17/c28-6-14-17(33)20(36)22(38)26(42-14)41-12-2-1-8(3-10(12)31)24-25(19(35)16-11(32)4-9(30)5-13(16)40-24)44-27-23(39)21(37)18(34)15(7-29)43-27/h1-5,14-15,17-18,20-23,26-34,36-39H,6-7H2/t14-,15-,17-,18-,20+,21+,22-,23-,26-,27+/m1/s1

InChI key

RPVIQWDFJPYNJM-DEFKTLOSSA-N

General description

Quercetin is an important flavonoid present in most of the vegetables including onion. Quercetin 3-glucuronide is one of the polyphenol metabolites, that is found to be localized to the brain.

Biochem/physiol Actions

Natural compound isolated from onions. Belongs to the class of organic compounds known as flavanoid-3-o-glycosides. Quercetin possesses free radical scavenging activity, due to the presence of phenolic hydroxyl groups present at the B-ring and position 3. The glycosidic form of quercetin is metabolized to its conjugates (glucuronide or sulfate) for absorption in the intestine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Multitargeted cancer prevention by quercetin
Murakami A, et al.
Cancer Letters, 269(2) (2008)
Antioxidant capacity of albumin-bound quercetin metabolites after onion consumption in humans
Murota K, et al.
The Journal of Medical Investigation, 54(3, 4) (2007)
Lap Ho et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(2), 769-781 (2012-10-26)
Epidemiological and preclinical studies indicate that polyphenol intake from moderate consumption of red wines may lower the relative risk for developing Alzheimer's disease (AD) dementia. There is limited information regarding the specific biological activities and cellular and molecular mechanisms by
Arshad Mehmood et al.
Combinatorial chemistry & high throughput screening, 23(9), 917-930 (2020-04-29)
The present study was designed to evaluate the xanthine oxidase (XO) inhibitory and antioxidant activities of 30 bioactive compounds present in edible food plants for the possible treatment of hyperuricemia. The XO inhibitory, SO and DPPH radical scavenging activities of
Enkhtaivan Gansukh et al.
Food chemistry, 334, 127508-127508 (2020-07-28)
Quercetin, a potential polyphenolic which possesses several biological effects. The influenza virus polymerase basic 2 (PB2) subunit of RNA polymerase responsible for replication, degree of virus conservation and active target site for designing specific antivirals. The quercetin derivatives downloaded from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service