Skip to Content
Merck
All Photos(1)

Documents

SMB00393

Sigma-Aldrich

β-GalNAc-PEG3-Azide

≥95% (HPLC)

Synonym(s):

(8-Azido-3,6-dioxaoctyl) 2-acetamido-2-deoxy-β-D-galactopyranoside, 2-{2-[2-azidoethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-β-D-galactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26N4O8
Molecular Weight:
378.38
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OCCOCCOCCN=[N+]=[N-])[C@H](NC(C)=O)[C@@H](O)[C@H]1O

InChI

1S/C14H26N4O8/c1-9(20)17-11-13(22)12(21)10(8-19)26-14(11)25-7-6-24-5-4-23-3-2-16-18-15/h10-14,19,21-22H,2-8H2,1H3,(H,17,20)/t10?,11?,12-,13+,14+/m0/s1

InChI key

FLYIOTWDNWJXHY-PNKMZZHYSA-N

Biochem/physiol Actions

β-GalNAc-PEG3-Azide is a PEGylated glycoside. PEGylated glycosides are used as ligands for Click Chemistry. They may also be important as carbohydrate carriers in glycotherapeutic applications, drug carriers for site-specific delivery, drug enhancers for decreasing toxicity, and solubility and protein enhancers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Eugenia Giorgi et al.
Beilstein journal of organic chemistry, 10, 1433-1444 (2014-07-06)
Conjugation with polyethylene glycol (PEG), known as PEGylation, has been widely used to improve the bioavailability of proteins and low molecular weight drugs. The covalent conjugation of PEG to the carbohydrate moiety of a protein has been mainly used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service