Skip to Content
Merck
All Photos(1)

Key Documents

SMB00010

Sigma-Aldrich

Albocycline

≥95% (LC/MS-ELSD)

Synonym(s):

Albocyclin, Cineromycin-β-methylaether, Ingramycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28O4
CAS Number:
Molecular Weight:
308.41
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C18H28O4/c1-13-7-6-8-14(2)16(21-5)9-11-18(4,20)12-10-17(19)22-15(13)3/h8-13,15-16,20H,6-7H2,1-5H3/b11-9+,12-10+,14-8+/t13-,15+,16-,18+/m0/s1

InChI key

BYWWNDLILWPPJP-REXWONOSSA-N

General description

Natural product derived from fungal source.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new inhibitor of melanogenesis, albocycline K3, produced by Streptomyces sp. OH-3984.
S Takamatsu et al.
The Journal of antibiotics, 49(5), 485-486 (1996-05-01)
Biosynthesis of albocycline: origin of the carbon skeleton.
A Taddei et al.
The Journal of antibiotics, 50(6), 526-528 (1997-06-01)
Albocycline- and carbomycin-type macrolides, inhibitors of human prolyl endopeptidases.
C Christner et al.
The Journal of antibiotics, 51(3), 368-371 (1998-05-20)
R C Thomas et al.
The Journal of antibiotics, 35(12), 1658-1664 (1982-12-01)
The structure and absolute configuration of the macrolide antibiotic albocycline (1a) has been determined by X-ray crystallographic analysis on the derived p-bromobenzoate (1b). The absolute configuration of albocycline is 4R, 7S, 12S, 13R.
K Harada et al.
The Journal of antibiotics, 37(10), 1187-1197 (1984-10-01)
As an approach to the search for new potentially useful macrolide antibiotics, we explored the minor components of albocycline (ALB) from the culture broth of Streptomyces bruneogriseus. Eight minor components were isolated and their structures were confirmed as 1 approximately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service