Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700005

Sigma-Aldrich

Anti-Fas Antibody

mouse monoclonal, LT95

Synonym(s):

Anti-FAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

product name

Monoclonal Anti-CD95 antibody produced in mouse, clone LT95, purified immunoglobulin, buffered aqueous solution

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LT95, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FAS(355)

Related Categories

General description

CD95, also known as Fas (fatty acid synthase) cell surface death receptor/APO-1, is encoded by the gene mapped to human chromosome 10q23.31. The encoded protein belongs to the tumor necrosis factor receptor superfamily and is expressed on a variety of cells including activated T and B cells. CD95 contains a death domain.
The antibody LT95 reacts with CD95 (Fas/APO-1), a 46 kDa single chain type I glycoprotein of the tumour necrosis factor/nerve growth factor (TNF/NGF) receptor superfamily, expressed on a variety of normal and neoplastic cells. It seems that the antibody LT95 does not induce Fas mediated apoptosis, although it cross-blocks anti-Fas DX2 antibody that recognizes a functional epitope of Fas molecule.

Immunogen

HUT-78 human T cell lymphoma cell line

Application

Monoclonal Anti-CD95 antibody produced in mouse has been used as a apoptosis inducer.

Biochem/physiol Actions

CD95/ Fas (fatty acid synthase) cell surface death receptor interacts with its natural ligand (FasL/CD95 L) and plays a vital role in regulation of T-cell homeostasis, by promoting apoptosis via activation-induced cell death (AICD). Mutation in the gene has been associated with the development of Sézary syndrome (SS) and Canale–Smith syndrome. CD95 gene loss can be used as a genetic marker of PSA recurrence after radical prostatectomy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in Tris buffered saline, pH 8.0, with 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chlorpyrifos and Cypermethrin Induce Apoptosis in Human Neuroblastoma Cell Line SH-SY5Y
Raszewski G
Basic and Clinical Pharmacology and Toxicology, 116, 158-167 (2015)
Downregulation of Fas Gene Expression in Sezary Syndrome Is Associated with Promoter Hypermethylation
Jones CL
The Journal of Investigative Dermatology, 130, 1116-1125 (2010)
Exploring prostate cancer genome reveals simultaneous losses of PTEN, FAS and PAPSS2 in patients with PSA recurrence after radical prostatectomy.
Ibeawuchi C
International Journal of Molecular Sciences, 16, 3856-3869 (2015)
Cytotoxicity-dependent APO-1 (Fas/CD95)-associated proteins form a death-inducing signaling complex (DISC) with the receptor.
Kischkel FC
The Embo Journal, 14, 5579-5588 (1995)
Meng Li et al.
Cancer cell, 35(6), 916-931 (2019-06-12)
Diffuse large B cell lymphomas (DLBCLs) are genetically heterogeneous and highly proliferative neoplasms derived from germinal center (GC) B cells. Here, we show that DLBCLs are dependent on mitochondrial lysine deacetylase SIRT3 for proliferation, survival, self-renewal, and tumor growth in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service