Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504327

Sigma-Aldrich

Anti-phospho-Werner Syndrome Helicase (pSer1141) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

RECQ3, RECQL2, WRN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 162 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer1141)

Gene Information

human ... WRN(7486)

Related Categories

General description

Werner syndrome, RecQ helicase-like (WRN) protein belongs to the RecQ family of DNA helicases. It has an adenosine triphosphate (ATP) binding site, one DExH helicase box, one exonuclease domain, and a nuclear localization signal. It is predominantly located in the nucleus. The WRN gene is located on human chromosome 8p12.

Immunogen

The antiserum was produced against synthesized peptide derived from human Werner Syndrome Helicase around the phosphorylation site of Ser1141.

Immunogen Range: 1107-1156

Biochem/physiol Actions

Werner syndrome, RecQ helicase-like (WRN) can interact with DNA polymerase δ (POLD1) and replication protein A (RPA) that are essential for the replication and repair of DNA. It also interacts with Ku protein which participates in double-strand DNA break repair. Mutation of the WRN gene results in Werner syndrome. WRN is a mortal target for cancer cells with microsatellite instability (MSI).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niek van Wietmarschen et al.
Nature, 586(7828), 292-298 (2020-10-02)
The RecQ DNA helicase WRN is a synthetic lethal target for cancer cells with microsatellite instability (MSI), a form of genetic hypermutability that arises from impaired mismatch repair1-4. Depletion of WRN induces widespread DNA double-strand breaks in MSI cells, leading
WRN (Werner syndrome, RecQ helicase-like)
Amor-Gueret M
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service