Skip to Content
Merck
All Photos(2)

Documents

SAB4501942

Sigma-Aldrich

Anti-CFTR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ABCC7, Cystic fibrosis transmembrane conductance regulator, cAMP- dependent chloride channel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 168 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CFTR(1080)

General description

Anti-CFTR Antibody detects endogenous levels of total CFTR protein.
CFTR (Cystic fibrosis transmembrane conductance regulator, ATP-binding cassette sub-family C, member 7) is a membrane-associated, N-linked glycoprotein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CFTR.

Immunogen Range: 711-760

Application

Anti-CFTR antibody produced in rabbit is suitable for immunohistochemistry and western blot applications.

Biochem/physiol Actions

CFTR (Cystic fibrosis transmembrane conductance regulator, ATP-binding cassette sub-family C, member 7) is mainly involved in the regulation of Na+ and Cl- transport by acting as a linear, cAMP activated, chloride channel. In addition, it is also associated with different transport signaling pathways. It has been reported that CFTR controls functionality of outwardly rectifying Cl- channels (ORCCs) by facilitating the transport and delivery of potent autacoid agonist and ORCC regulator ATP. It has also been suggested that CFTR can interact with Na+-reabsorptive pathway. CFTR is associated with congenital bilateral absence of the vas deferens (CBAVD) and causes the genital form of cystic fibrosis (CF). The CFTR gene may also responsible for male infertility. It may be useful for assisting reproduction technology.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S H Cheng et al.
Cell, 63(4), 827-834 (1990-11-16)
The gene associated with cystic fibrosis (CF) encodes a membrane-associated, N-linked glycoprotein called CFTR. Mutations were introduced into CFTR at residues known to be altered in CF chromosomes and in residues believed to play a role in its function. Examination
E M Schwiebert et al.
Cell, 81(7), 1063-1073 (1995-06-30)
The cystic fibrosis transmembrane conductance regulator (CFTR) functions to regulate both Cl- and Na+ conductive pathways; however, the cellular mechanisms whereby CFTR acts as a conductance regulator are unknown. CFTR and outwardly rectifying Cl- channels (ORCCs) are distinct channels but
Himanshu Sharma et al.
Gene, 548(1), 43-47 (2014-07-11)
High incidence of mutations in the cystic fibrosis transmembrane conductance regulator (CFTR) gene is associated with congenital bilateral absence of the vas deferens (CBAVD) and is considered as the genital form of cystic fibrosis (CF). The CFTR gene may also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service