Skip to Content
Merck
All Photos(3)

Key Documents

SAB4301791

Sigma-Aldrich

Anti-HTR2A

affinity isolated antibody

Synonym(s):

5-HT2A, HTR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat, human

technique(s)

immunohistochemistry: 1:20-1:100
western blot: 1:200-1:1000

accession no.

NP_000612

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTR2A(3356)

Immunogen

Synthetic peptide of human HTR2A

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dasiel O Borroto-Escuela et al.
ACS omega, 2(8), 4779-4789 (2017-09-19)
Studies on serotonin-selective reuptake inhibitors have established that disturbances in the ascending 5-HT neuron systems and their 5-HT receptor subtypes and collateral networks to the forebrain contribute to the etiology of major depression and are targets for treatment. The therapeutic
Barbara Chruścicka et al.
ACS chemical neuroscience, 10(7), 3225-3240 (2019-05-01)
The oxytocin receptor (OTR) and the 5-hydroxytryptamine 2A receptor (5-HTR2A) are expressed in similar brain regions modulating central pathways critical for social and cognition-related behaviors. Signaling crosstalk between their endogenous ligands, oxytocin (OT) and serotonin (5-hydroxytryptamine, 5-HT), highlights the complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service