Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200762

Sigma-Aldrich

Anti-Opsin antibody, Mouse monoclonal

clone RET-P1, purified from hybridoma cell culture

Synonym(s):

Anti-Opsin 2, Anti-Rhodopsin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

RET-P1, monoclonal

form

buffered aqueous solution

mol wt

~39 kDa

species reactivity

avian, bovine, fish, mouse, rat, human, amphibian, turtle, rabbit

concentration

~1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: 2-4 μg/mL using human retinoblastoma Y79 cell line extract.
immunofluorescence: suitable
immunohistochemistry: 5-10 μg/mL using rat eye frozen sections.
radioimmunoassay: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Rho(24717)

General description

Anti-Opsin antibody, Mouse monoclonal (rhodopsin) (mouse IgG1 isotype) is derived from the RET-P1 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/C mice immunized with rat retinal membranes. Opsin is also called as rhodopsin protein, members of the G-protein coupled receptor (GPCR) superfamily. Opsin comprises >95% of the rod outer segments (ROS) intrinsic membrane and consists of a protein moiety - an opsin - and a non-protein moiety - the chromophore retinal.

Application

Anti-Opsin antibody has been used in
  • immunoblotting
  • immunofluorescence
  • immunohistochemistry
  • enzyme linked immunosorbent assay (ELISA)
  • radioimmuno assay (RIA)

Biochem/physiol Actions

Opsin is an essential molecules for mediating the ability of animals to detect and use light for diverse biological functions. Mutations in the opsin (rhodopsin) gene are linked to retinitis pigmentosa (RP), a disease characterized by retinal degeneration resulting in reduced peripheral vision and night blindness.

Other Notes

rat retinal membranes

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The opsins
Terakita A.
Genome Biology, 6(3), 213-213 (2005)
Retinitis pigmentosa
Hamel C.
Orphanet Journal of Rare Diseases, 1(1), 40-40 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service