Skip to Content
Merck
All Photos(1)

Key Documents

SAB3701450

Sigma-Aldrich

Monoclonal Anti-Keratin antibody produced in mouse

clone C11, purified immunoglobulin

Synonym(s):

Anti-Non-histone chromosomal protein HMG-14 antibody, Anti-Type II keratin isoform K6c antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT6C(286887)

Specificity

This protein A purified mouse monoclonal antibody reacts specifically with keratins from human tissues and derived cell lines. This antibody reacts with keratin (56 kDa), keratin 17 (46 kDa), keratin 18 (45 kDa) and keratin 19 (40 kDa) derived from humans.  Cross reactivity with keratins from other sources has not been determined.  No reaction is expected against other filament proteins including vimentin, desmin and neurofilament protein.

Immunogen

Purified human cytoskeletal preparations from A431 cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% (w/v) Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniël Splinter et al.
PLoS biology, 8(4), e1000350-e1000350 (2010-04-14)
BICD2 is one of the two mammalian homologues of the Drosophila Bicaudal D, an evolutionarily conserved adaptor between microtubule motors and their cargo that was previously shown to link vesicles and mRNP complexes to the dynein motor. Here, we identified
Christine M Field et al.
Current biology : CB, 29(12), 1999-2008 (2019-06-11)
The large length scale of Xenopus laevis eggs facilitates observation of bulk cytoplasm dynamics far from the cortex during cytokinesis. The first furrow ingresses through the egg midplane, which is demarcated by chromosomal passenger complex (CPC) localized on microtubule bundles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service