Skip to Content
Merck
All Photos(2)

Key Documents

SAB2102351

Sigma-Aldrich

Anti-SYNJ1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-INPP5G, Anti-Synaptojanin 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

143 kDa

species reactivity

human, rabbit, pig, horse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYNJ1(8867)

Immunogen

Synthetic peptide directed towards the N terminal region of human SYNJ1

Biochem/physiol Actions

Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4,5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.Synaptojanin-1 is a phosphatidylinositol (4,5)-bisphosphate (PI(4,5)P2) that has a role in clathrin-coated pit dynamics Perera et al. (2006) [PubMed 17158794].[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-3383 AB020717.2 1-3383 3384-3539 AF009040.1 3318-3473 3540-4183 AF009039.1 3522-4165 4184-7039 AB020717.2 4142-6997

Sequence

Synthetic peptide located within the following region: IDSSDEDRISEVRKVLNSGNFYFAWSASGISLDLSLNAHRSMQEQTTDNR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Shi et al.
Microbiology spectrum, 12(4), e0200623-e0200623 (2024-02-15)
The gram-positive bacterium Staphylococcus aureus can invade non-professional phagocytic cells by associating with the plasma protein fibronectin to exploit host cell integrins. Integrin-mediated internalization of these pathogens is facilitated by the local production of phosphatidylinositol-4,5-bisphosphate (PI-4,5-P2) via an integrin-associated isoform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service