Skip to Content
Merck
All Photos(1)

Key Documents

S3322

Sigma-Aldrich

Supercinnamaldehyde

≥98% (HPLC)

Synonym(s):

1,3-Dihydro-1-methyl-3-(2-oxopropylidene)-2H-Indol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO2
CAS Number:
Molecular Weight:
201.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

faintly red to very dark red

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1C(=O)C(=C\C(C)=O)\c2ccccc12

InChI

1S/C12H11NO2/c1-8(14)7-10-9-5-3-4-6-11(9)13(2)12(10)15/h3-7H,1-2H3/b10-7+

InChI key

CZKBLHCEDVWPRN-JXMROGBWSA-N

Application

Supercinnamaldehyde may be used in transient receptor potential ankyrin 1 (TRPA 1)-mediated cell signaling studies.

Biochem/physiol Actions

Supercinnamaldehyde is a transient receptor potential ankyrin 1 (TRPA1) activator. EC50 = 0.8 μM.
Supercinnamaldehyde is a transient receptor potential ankyrin 1 (TRPA1) activator; EC50 = 0.8 μM; derivative of cinnamaldehyde; covalently binds to and activates TRPA1 receptor (expressed in nociceptive neurons) by modifying its cysteine residues. Covalent modification of reactive cysteines within TRPA1 can cause channel activation, rapidly signaling potential tissue damage through the pain pathway.

Caution

Air-sensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to transient receptor potential channels for your research needs.

We offer many products related to transient receptor potential channels for your research needs.

We offer many products related to transient receptor potential channels for your research needs.

We offer many products related to transient receptor potential channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service