Skip to Content
Merck
All Photos(2)

Key Documents

RAB0393

Sigma-Aldrich

Human Oncostatin M (OSM) ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1.37-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... OSM(5008)

General description

Oncostatin M (OSM) is a multifunctional hematopoietic cytokine that belongs to the interleukin-6 (IL-6) cytokine family. Human OSM is produced by activated T lymphocytes and monocytes mainly during inflammatory disorders. OSM acts as a cell growth modulator. In addition, it also participates in low-density lipoprotein (LDL) receptor upregulation and induction of hematopoietic factors. OSM contributes to the inflammatory process, autoimmunity, and immunopathogenesis of cancers.

Immunogen

Recombinant Human OSM

Application

The Human Oncostatin M (OSM) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Oncostatin M in serum, plasma, cell culture supernatants, and urine.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Grenier et al.
Blood, 93(4), 1413-1421 (1999-02-09)
Oncostatin M (OSM) is an interleukin-6 (IL-6) family cytokine known in particular to induce the synthesis of acute-phase proteins by hepatocytes. Because human polymorphonuclear neutrophils (PMN) can secrete numerous cytokines, the potential production of OSM by PMN was investigated. Highly
Y Ma et al.
Nucleic acids research, 27(23), 4649-4657 (1999-11-11)
Oncostatin M (OSM), an IL-6 subfamily cytokine, inhibits proliferation and causes morphological changes in many tumor cell lines. GM-CSF, phorbol-12-myristate-13-acetate (PMA), and lipopolysaccharide (LPS) induce OSM expression. To investigate the mechanisms governing OSM promoter activity, we have cloned and partially
Oncostatin M: A mysterious cytokine in cancers.
Masjedi, et al.
International Immunopharmacology, 90, 107158-107158 (2021)
Pulmonary expression of oncostatin M (OSM) promotes inducible BALT formation independently of IL-6, despite a role for IL-6 in OSM-driven pulmonary inflammation.
Botelho FM
Journal of Immunology, 191, 1453-1464 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service