Skip to Content
Merck
All Photos(3)

Key Documents

R0781

Sigma-Aldrich

Anti-Rab14 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-F protein-binding protein 1, Anti-GTPase Rab14, Anti-RAB14, member RAS oncogene family, Anti-Small GTP binding proein RAB14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~24 kDa

species reactivity

rat, human, mouse

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5 μg/mL using rat NRK cells fixed and permeabilized with 4% paraformaldehyde followed by 0.4% saponin
western blot: 2-4 μg/mL using whole extracts of mouse brain and human Raji cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB14(51552)
mouse ... Rab14(68365)
rat ... Rab14(94197)

General description

Ras-related protein Rab-14 (Rab14) is a member of the Rab11 subfamily of small guanosine triphosphatases (GTPases). It is associated with the early endosomal clathrin-coated trans‐Golgi network microdomain and is ubiquitously expressed. Rab14 belongs to the RAS oncogene family and is mapped to human chromosome 9q33.2.

Specificity

Anti-Rab14 recognizes human, rat, and mouse Rab14 (not tested in other species).

Application

Anti-Rab14 antibody produced in rabbit has been used in western blotting and immunofluorescence.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Ras-related protein Rab-14 (Rab14) is associated with the trafficking of the fibroblast growth factor receptor 2 (FGFR2) and regulates N-cadherin turnover. It is involved in membrane trafficking between the Golgi complex and endosomes. In polarized epithelial cells, Rab 14 regulates delivery of cargo from the trans‐Golgi network (TGN) to the apical membrane domain. Rab14 has also been implicated in phagosomal biogenesis. It acts as a tumor oncogene in non-small cell lung carcinoma (NSCLC) and is upregulated in gastric cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason T Schott et al.
Journal of visualized experiments : JoVE, (108), 53764-53764 (2016-03-12)
Efficient oligodendrogenesis is the therapeutic goal of a number of areas of research including spinal cord injury, neonatal hypoxia, and demyelinating diseases such as multiple sclerosis and transverse myelitis. Myelination is required to not only facilitate rapid impulse propagation within
Ella J Daly et al.
Neuropsychiatric disease and treatment, 3(6), 855-867 (2007-12-01)
Atypical antipsychotics are increasingly used in the treatment of a broad spectrum of psychiatric disorders. There is evidence that in addition to treating the positive and negative symptoms of schizophrenia, as well as mania in bipolar disorder, these agents may
Yu Chen et al.
The Journal of cell biology, 198(4), 545-560 (2012-08-22)
Rab proteins are important regulators of insulin-stimulated GLUT4 translocation to the plasma membrane (PM), but the precise steps in GLUT4 trafficking modulated by particular Rab proteins remain unclear. Here, we systematically investigate the involvement of Rab proteins in GLUT4 trafficking
D E Feierman et al.
The Biochemical journal, 239(3), 671-677 (1986-11-01)
Pyrazole and 4-methylpyrazole, which are inhibitors of alcohol dehydrogenase, were also found to be effective inhibitors of the oxidation of ethanol by liver microsomes (microsomal fractions) in vitro. Ethanol oxidation by microsomes from rats previously treated for 2 or 3
Khameeka N Kitt et al.
Traffic (Copenhagen, Denmark), 9(7), 1218-1231 (2008-04-24)
Epithelial cells display distinct apical and basolateral membrane domains, and maintenance of this asymmetry is essential to the function of epithelial tissues. Polarized delivery of apical and basolateral membrane proteins from the trans Golgi network (TGN) and/or endosomes to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service