Skip to Content
Merck
All Photos(2)

Key Documents

Q25120

Sigma-Aldrich

QAE Sephadex® A-25 chloride form

Synonym(s):

Diethyl-(2-hydroxypropyl)aminoethyl Sephadex®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

bead size

40-125 μm (dry)

Quality Level

pore size

~30,000 Da exclusion limit

operating pH

2-10

capacity

2.6-3.4 meq/g ion exchange capacity

compatibility

mode of use strong anion exchange chromatography

General description

Q25120-100G′s updated product number is GE17-0190-01

Application

QAE Sephadex® is used in protein chromatography, ion exchange chromatography, anion exchange media, resins and separation media. QAE Sephadex® has been used to develop methods to detect copper in water samples, resveratrol in beer, and indomethacin in both pharmaceuticals and urine samples.

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Varki et al.
Archives of biochemistry and biophysics, 222(1), 145-149 (1983-04-01)
An enzyme that is capable of removing the outer N-acetylglucosamine residues from phosphodiesters present on the high-mannose-type oligosaccharides of newly synthesized lysosomal enzymes has been described. This enzyme has been called an alpha-N-acetylglucosaminylphosphodiesterase, based upon its substrate specificity and on
3-deoxy-d-manno-2-octulosonic acid (KDO) is a component of rhamnogalacturonan II, a pectic polysaccharide in the primary cell walls of plants.
York, W.S., et al.
Carbohydrate Research, 138(1), 109-126 (1985)
A B Cantor et al.
The Journal of biological chemistry, 267(32), 23349-23356 (1992-11-15)
Cathepsin D is a bilobed lysosomal aspartyl protease that contains one Asn-linked oligosaccharide/lobe. Each lobe also contains protein determinants that serve as recognition domains for binding of UDP-GlcNAc:lysosomal enzyme N-acetylglucosamine-1-phosphotransferase, the first enzyme in the biosynthesis of the mannose 6-phosphate
Purification and Partial Characterization of Host-Specific Toxins Produced by <I>Periconia circinata</I>.
Wolpert, T.J., and Dunkle, L.D.
Phytopathology, 70, 872-876 (1980)
Heterogeneous Reaction of Shattercane to <I>Periconia circinata</I> and its Host-Specific Toxin.
Dunkle, L.D.
Phytopathology, 69, 260-262 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service