Skip to Content
Merck
All Photos(1)

Key Documents

PNS1010

Sigma-Aldrich

Stabilyser Reagent

for stabilization of DNA, RNA, and protein in tissue extracts

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
NACRES:
NA.77

form

liquid

Quality Level

feature

for stabilization of DNA, RNA, and protein in tissue extracts

shipped in

wet ice

storage temp.

2-8°C

General description

The stability of protein and antibody structure and function is dependent on a variety of factors such as temperature, freeze/thaw cycles, proteases and pH. For some proteins and antibodies, function can be lost completely with a single freeze/thaw cycle and proteases can significantly degrade proteins and antibodies within hours even when kept at 4 °C. Stabilyser Reagent minimizes the risk to improve protein assay quality and accuracy.

Stabilyser Reagent is a proprietary formulation of detergents and solutes optimized for efficient extraction, stabilization and storage of protein, DNA, and RNA from tissue samples. The unique composition of Stabilyser enables isolation of functionally active proteins, while maintaining integrity of nucleic acids. Stabilization of proteins and RNA is achieved even after 24 hours at room temperature Stabilyser reagent is compatible with Western blotting protocols, functional protein activity assays, and nucleic acid applications like qPCR.

Use in conjunction with a homognenizer for simultaneous tissue lysis and stabilization of protein and nucleic acids in tissue samples.

Application

Prevents degradation of proteins and nucleic acid during tissue lysis and storage. Stabilize functional proteins and RNA.

Features and Benefits

  • Gentle, non-denaturing lysis buffer does not interfere with downstream applications
  • Convenient ready-to-use reagent
  • Stabilizes RNA and protein for analysis at a later date

Preparation Note

Ready-to-use reagent. In general it is recommended to start optimization with 20 μL of Stabilyser reagent per mg of tissue.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service