Skip to Content
Merck
All Photos(2)

Key Documents

PLA0169

Sigma-Aldrich

Rabbit anti-NEK9 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

DKFZp434D0935, MGC16714, NERCC, NERCC1, NIMA (never in mitosis gene a)- related kinase 9, NIMA-related kinase 9, NIMA-related kinase Nek8, Nek8, nercc1 kinase, never in mitosis A-related kinase 9, nimA-related kinase 8, nimA-related protein kinase 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_149107.3

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... NEK9(91754)

Immunogen

The epitope recognized by PLA0169 maps to a region between residue 929 and 979 of human NIMA related kinase 9 using the numbering given in entry NP_149107.3 (GeneID 91754).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

NEK9 is a member of the NIMA-related serine/threonine kinases that play a role in mitotic progression. NEK9 plays a specific role in spindle organization and is necessary for accurate chromosome segregation. NEK9 may cooperate with the Ran-GTPase to regulate spindle formation. NEK6 and NEK7 are downstream substrates of NEK9 and may function cooperatively with NEK9 in mitotic spindle formation and maintenance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service