Skip to Content
Merck
All Photos(3)

Key Documents

PLA0081

Sigma-Aldrich

Rabbit anti-HIF1-alpha Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ARNT interacting protein, ARNT-interacting protein, HIF-1-alpha, HIF-1A, HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PAS domain-containing protein 8, PASD8, alpha subunit (basic helix-loop-helix transcription factor), bHLHe78, basic-helix-loop-helix-PAS protein MOP1, class E basic helix-loop-helix protein 78, hypoxia inducible factor 1, hypoxia-inducible factor 1, hypoxia-inducible factor 1 alpha isoform I.3, hypoxia-inducible factor1alpha, member of PAS protein 1, member of PAS superfamily 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

immunocytochemistry: 1:100-1:500
immunohistochemistry: 1:100-1:500
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

Q16665

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Immunogen

The epitope recognized by PLA0081 maps to a region between residues 775 and the C-terminus (residue 826) of human hypoxia-inducible factor 1, alpha subunit using the numbering given in Swiss-Prot entry Q16665 (GeneID 3091).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Hypoxia-inducible factor-1 (HIF1) is a basic helix-loop-helix (bHLH)-PER-ARNT-sim (PAS) transcription factor that plays a role in the cellular response to hypoxia. HIF1 is a heterodimer of an HIF1-alpha and HIF1-beta subunit. During normoxia, HIF-1alpha is regulated by ubiquitination and interaction with the von-Hippel-Lindau tumor suppressor protein (pVHL).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service