Skip to Content
Merck
All Photos(2)

Key Documents

NA0410

Sigma-Aldrich

GenElute HP Endotoxin-Free Plasmid Maxiprep Kit

greener alternative

sufficient for 25 preparations

Synonym(s):

Gen Elute, GenElute Endotoxin-Free Plasmid Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52
Pricing and availability is not currently available.

usage

sufficient for 25 preparations

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA extraction: suitable

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute HP Endotoxin-Free Plasmid Maxiprep Kit offers a simple and rapid method for isolating endotoxin-free plasmid DNA from recombinant E. coli cultures. The kit uses a vacuum format with a filter column for the rapid clearing of the bacterial lysate and a silica column for capturing plasmid DNA. A proprietary solution binds plasmid DNA to the binding column while preventing endotoxins from adsorbing. The technology allows for the user to consistently achieve levels of endotoxin that are less than 0.1 endotoxin units per μg of plasmid DNA.

High-quality, endotoxin-free DNA is ready for immediate use for the most demanding applications including transfection with endotoxin-sensitive cells.

Application

GenElute HP Endotoxin-Free Plasmid Maxiprep Kit has been used in the isolation of plasmid from swine liver[1] and from competent Escherichia coli cells.[2]

Features and Benefits

  • Up to 1.2 mg of high-copy plasmid DNA with endotoxin levels of ≤0.1 EU/μg
  • Only 35 minutes from pelleted cells to purified plasmid
  • Convenient vacuum format

Principle

An overnight recombinant E. coli culture is harvested by centrifugation and subjected to a modified alkaline-SDS lysis. The lysate is clarified by filtration followed by the addition of a binding solution that has been optimized for endotoxin-free plasmid preparations. The plasmid DNA is then captured on silica, while endotoxins are prevented from adsorbing to the membrane. Two wash steps remove contaminants. Finally, the bound DNA is eluted in endotoxin-free water. The recovered plasmid DNA is predominately in its supercoiled form. Genomic DNA and RNA are below detectable levels by ethidium bromide stained agarose gel electrophoresis.

Other Notes

For additional information, please see www.sigma-aldrich.com/genelutehp.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Successful liver-directed gene delivery by ERCP-guided hydrodynamic injection (with videos)
Kumbhari V, et al.
Gastrointestinal Endoscopy, 88(4), 755-763 (2018)
Towards a metalloprotease-DNA vaccine against piscine cryptobiosis caused by Cryptobia salmositica
Tan CW, et al.
Parasitology Research, 102(2), 265-275 (2008)
Keith Hansen et al.
Journal of visualized experiments : JoVE, (64)(64), e3304-e3304 (2012-06-27)
Genome editing is a powerful technique that can be used to elucidate gene function and the genetic basis of disease. Traditional gene editing methods such as chemical-based mutagenesis or random integration of DNA sequences confer indiscriminate genetic changes in an
João M Furtado et al.
Investigative ophthalmology & visual science, 53(11), 6856-6862 (2012-09-07)
Toxoplasma gondii, the parasite responsible for ocular toxoplasmosis, accesses the retina from the bloodstream. We investigated the dendritic cell as a potential taxi for T. gondii tachyzoites moving across the human retinal endothelium, and examined the participation of adhesion molecules
Paul A Beare et al.
Journal of bacteriology, 191(5), 1369-1381 (2008-12-31)
Coxiella burnetii is a gram-negative obligate intracellular bacterium and the causative agent of human Q fever. The lack of methods to genetically manipulate C. burnetii significantly impedes the study of this organism. We describe here the cloning and characterization of

Related Content

GenElute HP Plasmid Maxiprep Kit isolates endotoxin-free plasmid DNA in 35 minutes, meeting industry standards.

GenElute HP Plasmid Maxiprep Kit isolates endotoxin-free plasmid DNA in 35 minutes, meeting industry standards.

GenElute HP Plasmid Maxiprep Kit isolates endotoxin-free plasmid DNA in 35 minutes, meeting industry standards.

GenElute HP Plasmid Maxiprep Kit isolates endotoxin-free plasmid DNA in 35 minutes, meeting industry standards.

Questions

1–6 of 6 Questions  
  1. Which format gives a better yield, vacuum or spin?

    1 answer
    1. There is no difference in plasmid recovery when using either format.

      Helpful?

  2. Do the binding columns fit on other manufacturers’ manifolds?

    1 answer
    1. Yes. The columns fit on all standard vacuum manifolds.

      Helpful?

  3. What is the expected yield of plasmid DNA that you can isolate from these kits?

    1 answer
    1. Yields can reach as high as 350 mg for a midiprep and 1.2 mg for a maxiprep. As with any protocol, lower yields can be expected with low copy plasmids.

      Helpful?

  4. Is plasmid DNA isolated with this kit suitable for use in transfection?

    1 answer
    1. Yes. However, if you are currently required to use an endofree plasmid purification kit, we recommend using the GenElute HP Endotoxin-free Kit.

      Helpful?

  5. How do the GenElute HP Plasmid Kits differ from other plasmid purification systems?

    1 answer
    1. The GenElute HP Plasmid Kits are much faster than other commercially available kits. A filter syringe is used for preparing the cleared lysate, which replaces a lengthy centrifugation step. These kits also offer the option of using a vacuum or spin purification format rather than a gravity drip to bind, wash and elute the plasmid. In addition, samples are eluted in Tris buffer so there are no alcohol precipitations required.

      Helpful?

  6. How long does it take to isolate plasmid using the vacuum format?

    1 answer
    1. A midiprep or maxiprep requires only 30 minutes to complete.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service