MTCA8001R
MTC™ Bio BowTie™ Drawstring Biohazard Bags
high-density polyethylene, capacity 12 gal (45 L), W × L 25 in. (64 cm) × 35 in. (89 cm), with large marking area and sterilization indicator, pack of 100 ea
Synonym(s):
bio waste bag
All Photos(1)
About This Item
UNSPSC Code:
24111500
NACRES:
NB.15
Recommended Products
material
high-density polyethylene
packaging
pack of 100 ea
manufacturer/tradename
MTC™ Bio A8001R
W × L
25 in. (64 cm) × 35 in. (89 cm)
capacity
12 gal (45 L)
General description
Bowtie™ Biohazard bags, autoclave bag with drawstring, PE, 25 x 35in. (64 x 89cm), with marking area and sterilization indicator, 100/pk
Features and Benefits
- Drawstring for instant closure
- 12-16 gallon capacity, 25 x 35in.
- (45-60 liter capacity, 64 x 89cm)
- Bags fit perfect in Biohazard Bin
- Bin has a hands-free foot pedal operated lid
Legal Information
BowTie is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)