Skip to Content
Merck
All Photos(6)

Key Documents

M4819

Sigma-Aldrich

Anti-methyl-Histone H3 (Me-Lys4) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-H3K4me1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
Caenorhabditis elegans, rat, chicken, Drosophila, bovine, Arabidopsis thaliana, human, mouse, frog
citations:
14
technique(s):
western blot: 0.5-1 μg/mL using whole cell extract of human HeLa cells, human Jurkat cells, or mouse NIH3T3 cells

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 17 kDa

species reactivity

Caenorhabditis elegans, rat, chicken, Drosophila, bovine, Arabidopsis thaliana, human, mouse, frog

technique(s)

western blot: 0.5-1 μg/mL using whole cell extract of human HeLa cells, human Jurkat cells, or mouse NIH3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

monomethylation (Lys4)

General description

Histones are proteins that form the nucleosome along with DNA. A nucleosome consists of core histone octamers made of two heterodimers of H2A and H2B along with two heterodimers of H3 and H4. This octamer is bound by the DNA and H1 protein. The HIST3H3 gene is mapped on the human chromosome at 1q42.13.

Specificity

Anti-methyl-Histone H3 [Me-Lys4] recognizes histone H3 methylated on Lys4.

Immunogen

synthetic methylated peptide corresponding to amino acid 1-8 (Me-Lys4) at the N-terminus of human histone H3, conjugated to KLH. The sequence is identical in many species.

Application

Anti-methyl-Histone H3 (Me-Lys4) antibody produced in rabbit detection of [Me-Lys4] histone H3 by immunoblotting, chromatin immunoprecipitation and immunoprecipitation.

Biochem/physiol Actions

Histone methylation like acetylation is a complex dynamic process involved in several processes including transcriptional regulation, chromatin condensation, mitosis, and heterochromatin assembly. Histones H3 and H4 are the predominant histones modified by methylation and are highly methylated in mammalian cells. Lysine residues can be mono-, di-, and trimethylated at different heterochromatic subdomains adding further complexity to the regulation of chromatin structure. Conserved lysine residues in the N- terminal tail domains of histone H3 (Lys4, Lys9, and Lys27) are the preferred sites of methylation. Histone H3 mono-, di-, and trimethylation at Lys4 and Lys9 are carried out both in vitro and in vivo by SET domain site-specific histone methyltransferases (HMTases), including Suv39h1, Suv39h2, and G9a. Di- and trimethylation of histone H3 at Lys4 in coding regions correlates with transcriptional activity of many genes. Dimethylation of Histone H3 at Lys4 occurs at both active and inactive euchromatic regions, but not in silent heterochromatic sites, whereas trimethylation at Lys4 is present exclusively at active genes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.Sigma brand products are sold through Sigma-Aldrich, Inc.Sigma-Aldrich, Inc. warrants that its products conform to the information contained in this and other Sigma-Aldrich publications.Purchaser must determine the suitability of the product(s) for their particular use. Additional terms and conditions may apply. Please see reverse side of the invoice or packing slip.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Wang et al.
Journal of the American Chemical Society, 142(35), 15098-15106 (2020-08-17)
Enzyme assays are important for many applications including clinical diagnostics, functional proteomics, and drug discovery. Current methods for enzymatic activity measurement often suffer from low analytical sensitivity. We developed an ultrasensitive method for the detection of enzymatic activity using Single
Prostacyclin synthase expression and epigenetic regulation in nonsmall cell lung cancer
Cathcart MC, et al.
Cancer, 117(22), 5121-5132 (2011)
Epigenetics Underpinning the Regulation of the CXC (ELR+) Chemokines in Non-Small Cell Lung Cancer
Baird AM, et al.
PLoS ONE, 6(1), e14593-e14593 (2011)
IL-20 is epigenetically regulated in NSCLC and down regulates the expression of VEGF
Baird, Anne-Marie and others
European Journal of Cancer, 47(12), 1908-1918 (2011)
Helena Santos-Rosa et al.
Nature, 419(6905), 407-411 (2002-09-28)
Lysine methylation of histones in vivo occurs in three states: mono-, di- and tri-methyl. Histone H3 has been found to be di-methylated at lysine 4 (K4) in active euchromatic regions but not in silent heterochromatic sites. Here we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service