Skip to Content
Merck
All Photos(1)

Key Documents

L4537

Greiner multiwell plate lids

for 96 well plate with condensation rings, clear, sterile

Synonym(s):

microplate lids, microtiter plate lids, multiwell plate lids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
colorless wells
size:
96 well
sterility:
sterile
binding type:
non-treated surface
feature:
lid
Pricing and availability is not currently available.

material

colorless wells

sterility

sterile

feature

lid

packaging

pack of 5

manufacturer/tradename

Greiner 656176

size

96 well

color

clear

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Greiner multiwell plate lids are made of polystyrene.
  • Corner notches for proper orientation.
  • Robotics-friendly full- and Lo-profile skirt options.
  • Free of detectable RNase and DNase.
Corner notches for proper orientation. Robotics friendly full- and Lo-profile skirt options. RNase and DNase free.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service