Skip to Content
Merck
All Photos(1)

Key Documents

HUMANBMMC-0002115

Sigma-Aldrich

Frozen Human Normal Bone Marrow Mononuclear Cells (BMMCs)

human bone marrow (Mononuclear Cells)

Synonym(s):

BMMC, Bone Marrow Mononuclear Cells, Immune Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

Frozen Human Normal Bone Marrow Mononuclear Cells (BMMCs), 10 Million Cells/per vial

biological source

human bone marrow (Mononuclear Cells)

Quality Level

form

frozen liquid

packaging

vial of 10 Million cells

manufacturer/tradename

BioIVT

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


We are pleased to offer these products from BioIVT.

Bone Marrow Mononuclear Cells (BMMCs) are a heterogenous mixture of cells isolated from bone marrow and is especially rich in hematopoeitic stem cells. BMMCs consist of several types of cells including lymphocytes, monocytes, mesenchymal stem cells and hematopoietic progenitor cells and are isolated from bone marrow aspirate via density gradient centrifugation.

Used for a variety of applications, BMMCs are frequently utilized in drug discovery/development, assay validation/development, cell therapy research, and other immunology applications.

Packaging

10 Million Cells/per vial

Other Notes

Thaw before use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service