HS2860B
TrueNorth® Cardboard Cryovial Box and Lid
without dividers, for tubes, 1.5 ‑ 2 mL, L × W × H 5.2 × 5.2 × 3 in., white, pk of 12
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
cardboard
feature
for tubes 1.5 ‑ 2 mL
packaging
pk of 12
manufacturer/tradename
Heathrow Scientific HS2860B
technique(s)
cryopreservation: suitable
L × W × H
5.2 × 5.2 × 3 in.
size
133x133x75 mm
color
white
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- It stores small samples and tubes.
- Store in mechanical freezers or vapor phase LN2 at -196ºC.
- Use tubes that can be stored at the required temperature.
- Colors make it simple to identify and categorize items.
- Bottom vent holes enable efficient circulation and evacuation of cold air, vapor, or LN2.
- All box footprints are 13.3 x 13.3 cm, select from two different heights
- Autoclaving is not recommended.
Other Notes
Order partitions separately.
Legal Information
TrueNorth is a registered trademark of Heathrow Scientific LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service