Skip to Content
Merck
All Photos(3)

Key Documents

HPA035434

Sigma-Aldrich

Anti-MTHFD2L antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Mthfd2L Antibody, Mthfd2L Antibody - Anti-MTHFD2L antibody produced in rabbit, Anti-MGC72244, Anti-methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PASHTYVRNKIRAASAVGICSELILKPKDVSQEELLDVTDQLNMDPRVSGILVQLPLPDHVDERTI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein (MTHFD2L) is an adult mitochondrial MTHFD isozyme present in mammalian cells. The MTHFD2L gene is located on the human chromosome at 4q13.3.

Immunogen

methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-MTHFD2L antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein (MTHFD2L) catalyzes the conversion of 5,10-methylenetetrahydrofolate (CH2-THF) into formyl-THF (CHO-THF), a substrate to produce formate. It possesses bifunctional activities of CH2-THF dehydrogenase and 5,10-methenyl-THF cyclohydrolase.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79817

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combined donor and recipient non-HLA genotypes show evidence of genome-wide association with transplant-related mortality (TRM) after HLA-matched unrelated donor blood and marrow transplantation (URD-BMT)(DISCOVeRY-BMT study)
Sucheston-Campbell L E, et al.
Blood (2015)
Denise R Minton et al.
Molecular cell, 69(4), 610-621 (2018-02-18)
Upon glucose restriction, eukaryotic cells upregulate oxidative metabolism to maintain homeostasis. Using genetic screens, we find that the mitochondrial serine hydroxymethyltransferase (SHMT2) is required for robust mitochondrial oxygen consumption and low glucose proliferation. SHMT2 catalyzes the first step in mitochondrial
R Nilsson et al.
Gene, 716, 144032-144032 (2019-08-05)
Mitochondrial folate metabolism is central to the generation of nucleotides, fuelling methylation reactions, and redox homeostasis. Uniquely among the reactions of the mitochondrial folate pathway, the key step of the oxidation of 5,10-methylene-tetrahydrofolate (CH2-THF) can be catalysed by two isozymes
Minhye Shin et al.
The Journal of biological chemistry, 289(22), 15507-15517 (2014-04-16)
Mammalian mitochondria are able to produce formate from one-carbon donors such as serine, glycine, and sarcosine. This pathway relies on the mitochondrial pool of tetrahydrofolate (THF) and several folate-interconverting enzymes in the mitochondrial matrix. We recently identified MTHFD2L as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service