Skip to Content
Merck
All Photos(2)

Key Documents

H0392

Sigma-Aldrich

Hemoglobin S, Ferrous Stabilized human

lyophilized powder

Synonym(s):

Sickle cell hemoglobin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42030116
NACRES:
NA.61
Pricing and availability is not currently available.

biological source

human

Quality Level

Assay

97-100% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

electrophoresis: suitable
immunofluorescence: suitable

solubility

H2O: soluble 20 mg/mL

suitability

suitable for electrophoresis and chromatography standard

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HBB(3043)

Looking for similar products? Visit Product Comparison Guide

General description

Hemoglobin is the major component of red blood cells, and is responsible for their red color. Its normal concentration in erythrocytes is 34%. Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide.
has not been tested for functional equivalence against native preparations (unlyophilized ferrous hemoglobins).

Application

Hemoglobin S was used in the determination of fetal hemoglobin by time-resolved immunofluorometric assay.[1]

Packaging

Package size indicates the amount of hemoglobin as determined by the procedure of Drabkin, D.L., J. Biol. Chem., 164, 703 (1946).

Reconstitution

When reconstituted with buffer, gives >90% ferrous hemoglobin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian T Castle et al.
Science advances, 5(3), eaau1086-eaau1086 (2019-03-21)
In sickle cell disease, the aberrant assembly of hemoglobin fibers induces changes in red blood cell morphology and stiffness, which leads to downstream symptoms of the disease. Therefore, understanding of this assembly process will be important for the treatment of
U Turpeinen et al.
Clinical chemistry, 38(10), 2013-2018 (1992-10-01)
We have developed a "sandwich"-type time-resolved immunofluorometric assay (IFMA) for fetal hemoglobin (HbF) in hemolysates from adults and newborns, amniotic fluid, and plasma, based on a polyclonal and a monoclonal antibody against human fetal hemoglobin. Microtiter wells are coated with
Charles T Quinn et al.
British journal of haematology, 175(4), 724-732 (2016-09-09)
Sickle cell disease (SCD) is a significant healthcare burden worldwide, but most affected individuals reside in low-resource areas where access to diagnostic testing may be limited. We developed and validated a rapid, inexpensive, disposable diagnostic test, the HemoTypeSC™ , based

Questions

  1. How many tablets are in the bottle?

    1 answer
    1. This product is not packaged in tablets, it is a supplied in powdered format. It is currently offered in a 25 mg and 100 mg pack size.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service