Skip to Content
Merck
All Photos(1)

Key Documents

GERPN202

Anti-Brdu Antibody

Cytiva RPN202

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

conjugate

unconjugated

antibody product type

primary antibodies

packaging

pkg of 130 μL

manufacturer/tradename

Cytiva RPN202

storage temp.

2-8°C

General description

The Anti-Bromodeoxyuridine Antibody is supplied in the same formulation as the monoclonal antibody to BrdU in the kit. The antibody/nuclease mixture is compatible with detection systems that incorporate fluorescently labeled antibodies and biotin-streptavidin detection reagents.

Features and Benefits

  • The monoclonal antibody to Bromo-deoxyuridine is supplied in the.same formulation used in the Cytiva Cell Proliferation Kit (RPN20).
  • No need for chemical denaturation.
  • Detection while maintaining cell morphology.
  • Double immunocytochemical staining studies can be perfromed.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samir A Bello et al.
Journal of marine science and engineering, 10(2) (2022-10-27)
The cellular events underlying intestine regrowth in the sea cucumber Holothuria glaberrima have been described by our group. Currently, the molecular and signaling mechanisms involved in this process are being explored. One of the limitations to our investigations has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service