Key Documents
GE28-9323-60
Vivaspin® 20, 10 kDa MWCO Polyethersulfone
Cytiva 28-9323-60, pack of 12 pieces
About This Item
UNSPSC Code:
41104916
NACRES:
NB.24
Pricing and availability is not currently available.
material
polycarbonate housing
polyethersulfone membrane
sterility
non-sterile
packaging
pack of 12 pieces
manufacturer/tradename
Cytiva 28-9323-60
availability
not available in Japan
tube size
50 mL , conical bottom
working volume
20 mL
pore size
10 kDa MWCO
storage temp.
room temp
General description
Designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration. Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%.
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration. Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cut-off (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.
Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3 000 to 100 000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest
Features and Benefits
  • Compatible pH range is from pH 1 to 9.
  • Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss.
  • Vertical polyethersulfone membrane minimizes membrane blockage and tolerates high flow rates.
Storage and Stability
Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Vivaspin is a registered trademark of Sartorius Stedim Biotech GmbH
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
This page describes troubleshooting strategies in purification of GST-tagged proteins using Cytiva products.
Protocols
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
See All