GE28400737
Large Column Holder
For columns o.d. 50 mm, ÄKTA®xpress, Cytiva 28400737
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151816
NACRES:
NB.13
Recommended Products
feature
For columns o.d. 50 mm, ÄKTA®xpress
Connections: 5/16" male
Wetted Material: Polyetheretherketone (PEEK)
manufacturer/tradename
Cytiva 28400737
General description
Column holder for using columns with o.d. 50 mm together with ÄKTA®xpress.
Legal Information
ÄKTA is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Shamekh et al.
Journal of advances in modeling earth systems, 12(11), e2020MS002164-e2020MS002164 (2020-12-08)
This study investigates the feedbacks between an interactive sea surface temperature (SST) and the self-aggregation of deep convective clouds, using a cloud-resolving model in nonrotating radiative-convective equilibrium. The ocean is modeled as one layer slab with a temporally fixed mean
Zena Al-Bakri et al.
Regenerative therapy, 15, 53-63 (2021-01-12)
Neural crest (NC)-like stem/progenitor cells provide an attractive cell source for regenerative medicine because of their multipotent property and ease of isolation from adult tissue. Although human umbilical cord blood (HUCB) is known to be a rich source of stem
Lehti Saag et al.
Science advances, 7(4) (2021-02-02)
The transition from Stone to Bronze Age in Central and Western Europe was a period of major population movements originating from the Ponto-Caspian Steppe. Here, we report new genome-wide sequence data from 30 individuals north of this area, from the
Ruochi Zhang et al.
Cell systems, 10(5), 397-407 (2020-06-19)
Recent advances in ligation-free, genome-wide chromatin interaction mapping such as SPRITE and ChIA-Drop have enabled the identification of simultaneous interactions involving multiple genomic loci within the same nuclei, which are informative to delineate higher-order genome organization and gene regulation mechanisms
Yui Ishiguro et al.
Annals of surgical oncology, 28(2), 1178-1186 (2020-08-10)
Circulating tumor cells (CTCs) have been shown to be heterogeneous. Focusing on the epithelial-mesenchymal transition and perioperative kinetics, we evaluated CTCs with mesenchymal phenotypes as a potential prognostic biomarker for patients with gastric cancer. Peripheral blood was collected from 54
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service